Skip to Content
Merck
All Photos(3)

Documents

ECM515

Sigma-Aldrich

QCM Chemotaxis Cell Migration Assay, 96-well (3 µm), fluorimetric

The Chemicon QCM 3 uM 96-well Migration Assay provides a quick & efficient system for quantitative determination of various factors on cell migration, including screening of pharmacological agents or evaluation of integrins.

Synonym(s):

Chemotaxis Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®
QCM

technique(s)

activity assay: suitable
cell based assay: suitable

detection method

fluorometric

shipped in

wet ice

Application

Research Category
Cell Structure

Packaging

96 wells

Components

Sterile 96-well 3μM Cell Migration Plate Assembly: (Part No. 90199) One 96-well feeder tray, and one 96-well Cell Migration Chamber plate with 3 μm pore insert. Area = 0.3 cm2, 50,000 cells per well

96-well Cell Culture Tray: (Part No. 90129) One 96-well feeder tray.

Cell Detachment Solution: (Part No. 90131) One bottle - 16 mL.

4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL.

CyQuant GR Dye: (Part No. 90132) One vial - 75 μL

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Campylobacter jejuni invade chicken LMH cells inefficiently and stimulate differential expression of the chicken CXCLi1 and CXCLi2 cytokines.
Charles L Larson, Devendra H Shah, A Singh Dhillon, Douglas R Call, Soohyoun Ahn et al.
Microbiology (Reading, England) null
Jon M Davis et al.
Infection and immunity, 74(8), 4401-4408 (2006-07-25)
Cytotoxic necrotizing factor type 1 (CNF1), a toxin produced by many strains of uropathogenic Escherichia coli (UPEC), constitutively activates small GTPases of the Rho family by deamidating a single amino acid within these target proteins. Such activated GTPases not only
Clemente J Britto et al.
American journal of physiology. Lung cellular and molecular physiology, 316(2), L321-L333 (2018-11-22)
Bpifa1 (BPI fold-containing group A member 1) is an airway host-protective protein with immunomodulatory properties that binds to LPS and is regulated by infectious and inflammatory signals. Differential expression of Bpifa1 has been widely reported in lung disease, yet the
Danielle K Lewis et al.
Neurobiology of aging, 33(6), 1123-1123 (2011-12-14)
In middle-aged female rats, focal ischemia leads to a larger cortical infarction as compared with younger females. To determine if stroke-induced cytotoxicity in middle-aged females was associated with impaired astrocyte function, astrocytes were harvested and cultured from the ischemic cortex

Related Content

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service