Skip to Content
Merck
All Photos(1)

Key Documents

AX1385

Supelco

Ammonium sulfate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-984-1
Assay:
≥99.0%
form:
crystals
solubility:
754-764 g/L
Pricing and availability is not currently available.

Agency

meets USP/NF

Quality Level

vapor pressure

<1 Pa ( 25 °C)

Assay

≥99.0%

form

crystals

impurities

≤0.005% Insoluble matter

ign. residue

≤0.005%

color

white

pH

5 (100 g/L in H2O)

mp

>280 °C (dec.) (lit.)

solubility

754-764 g/L

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤5 ppm

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.0% minChloride (Cl)5ppm maxColorWhiteFormCrystalsHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.005% maxIron (Fe)5ppm maxNitrate (NO3)0.001% maxPhosphate (PO4)5ppm maxResidue after ignition0.005% maxpH of a 5% solution at 25°C5.0-6.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service