Skip to Content
Merck
All Photos(1)

Key Documents

AP160

Sigma-Aldrich

Rabbit Anti-Mouse IgG Antibody

2.4 mg/mL, Chemicon®

Synonym(s):

IgG antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

2.4 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Reacts with Mouse IgG gamma chain, as well as the light chains from all Mouse immunoglobulin classes. Monospecific on IEP against whole Mouse Serum.

Immunogen

Highly purified Mouse IgG isolated from pooled normal Mouse Serum.

Application

Detect Mouse IgG using this Rabbit anti-Mouse IgG Antibody validated for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Storage and Stability

Store at +2-8°C. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Collateral projection from the locus coeruleus to whisker-related sensory and motor brain regions of the rat.
Sat-Byol Lee,Suk K Beak,Seung H Park,Barry D Waterhouse,Hyun S Lee
The Journal of Comparative Neurology null
Jesús M López et al.
Brain, behavior and evolution, 90(4), 289-310 (2017-11-22)
The distribution of DARPP-32 (a phosphoprotein related to the dopamine D1 receptor) has been widely used as a means to clarify the brain regions with dopaminoceptive cells, primarily in representative species of tetrapods. The relationship between dopaminergic and dopaminoceptive elements
Immunohistochemical localization of DARPP-32 in the brain and spinal cord of anuran amphibians and its relation with the catecholaminergic system.
Jesus M Lopez,Ruth Morona,Agustin Gonzalez
Journal of Chemical Neuroanatomy null
Kari Vaahtomeri et al.
Journal of cell science, 121(Pt 21), 3531-3540 (2008-10-09)
Inactivating mutations of the tumor-suppressor kinase gene LKB1 underlie Peutz-Jeghers syndrome (PJS), which is characterized by gastrointestinal hamartomatous polyps with a prominent smooth-muscle and stromal component. Recently, it was noted that PJS-type polyps develop in mice in which Lkb1 deletion
Michael J Scherm et al.
Cell reports, 41(4), 111562-111562 (2022-10-27)
Infection by the Ebola virus, a member of the Filoviridae family of RNA viruses, leads to acute viral hemorrhagic fever. End-stage Ebola virus disease is characterized by a cytokine storm that causes tissue damage, vascular disintegration, and multi-organ failure. Previous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service