Skip to Content
Merck
All Photos(3)

Key Documents

ABS1004

Sigma-Aldrich

Anti-phospho-NKCC1 Antibody (Thr212/Thr217)

serum, from rabbit

Synonym(s):

phospho-NKCC1 Thr212/Thr217, Solute carrier family 12 member 2, Basolateral Na-K-Cl symporter, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr212/pThr217)

Gene Information

human ... SLC12A2(6558)

General description

The Na-K-Cl cotransporter 1 (NKCC1) is a membrane protein coded by the SLC12A2 gene and functions in cellular sodium/potassium/chloride transport and homeostasis. It is widely distributed throughout the body and serves important functions, especially in liquid-secreting glands. Phosphorylation of Thr212/Thr217 in the intracellular N-terminal domain of NKCC1 is correlated with NKCC1 activation.

Specificity

Specific for mature form of phosphorylated NKCC1.

Immunogen

Linear peptide corresponding to human phospho-NKCC1 Thr212/Thr217.

Application

Anti-phospho-NKCC1 Thr212/Thr217 is an antibody against phospho-NKCC1 for use in Western Blotting, Immunofluorescence.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected phospho-NKCC1 Thr212/Thr217 in GMB43 and GBM5 cell lysate (Dr. Anders Persson, UCSF) (Grim, R.P., et al. (2012). JBC. 1-31).
Immunofluorescence Analysis: A representative lot detected phospho-NKCC1 Thr212/Thr217 in GBM43 human tissiue (Dr. Anders Persson, UCSF) (Grim, R.P., et al. (2012). JBC. 1-31).

Quality

Evaluated by Western Blotting in GMB43 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-NKCC1 Thr212/Thr217 in 10 µg of GMB43 cell lysate.

Target description

~250 kDa observed. Likely a dimer of mature form of NKCC1, NKCC1 is glycosylated differently in different cells, leading to uncharacterized band(s) in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Kopecky et al.
Scientific reports, 12(1), 4609-4609 (2022-03-19)
Glioblastoma has remained the deadliest primary brain tumor while its current therapy offers only modest survival prolongation. Immunotherapy has failed to record notable benefits in routine glioblastoma treatment. Conventionally, immunotherapy relies on T cells as tumor-killing agents; however, T cells
Shamma S Rahman et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(12), 5217-5233 (2017-08-06)
The Eps15-homology domain-containing (EHD) protein family comprises 4 members that regulate endocytic recycling. Although the kidney expresses all 4 EHD proteins, their physiologic roles are largely unknown. This study focused on EHD4, which we found to be expressed differentially across
Chong Jiang et al.
The Journal of biological chemistry, 292(8), 3137-3145 (2017-01-15)
The ubiquitin ligase Nedd4-like (Nedd4L, or Nedd4-2) binds to and regulates stability of the epithelial Na+ channel (ENaC) in salt-absorbing epithelia in the kidney, lung, and other tissues. Its role in the distal colon, which also absorbs salt and fluid
Yang Chen et al.
Frontiers in cellular neuroscience, 17, 1279385-1279385 (2023-12-18)
Neuroinflammation is a pathological event associated with many neurological disorders, including dementia and stroke. The choroid plexus (ChP) is a key structure in the ventricles of the brain that secretes cerebrospinal fluid (CSF), forms a blood-CSF barrier, and responds to
Zhaoqi Zhang et al.
Journal of neuroinflammation, 19(1), 163-163 (2022-06-22)
Hydrocephalus is a severe complication of intracerebral hemorrhage with ventricular extension (ICH-IVH) and causes cerebrospinal fluid (CSF) accumulation. The choroid plexus epithelium plays an important role in CSF secretion and constitutes the blood-CSF barrier within the brain-immune system interface. Although

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service