Skip to Content
Merck
All Photos(3)

Key Documents

ABE413

Sigma-Aldrich

Anti-NF-E2-related factor 2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear factor erythroid 2-related factor 2, NF-E2-related factor 2, NFE2-related factor 2, HEBP1, Nuclear factor erythroid derived 2-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NFE2L2(4780)

Related Categories

General description

NF-E2-related factor 2 (Nuclear factor erythroid 2-related factor 2) is a transcription activator that binds to antioxidant response elements (ARE) in the promoter regions of target genes. NF-E2-related factor 2 mediates the coordinated up-regulation of genes in response to oxidative stress. NF-E2-related factor 2 may also be involved in the transcriptional activation of genes of the beta-globin cluster. NF-E2-related factor 2 is widely expressed, with highest expression in adult muscle, kidney, lung, liver and fetal muscle.

Specificity

This antibody recognizes the C-terminus of NF-E2 related factor 2

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human NF-E2 related factor 2.

Application

Anti-NF-E2-related factor 2 Antibody is a highly specific rabbit polyclonal antibody, that targets NF-E2-related factor 2 & has been tested in western blotting & ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western Blotting Analysis: 1 µg/mL from a representative lot detected NF-E2 related factor 2 in 10 µg of Jurkat cell lysate.

Immunocytochemistry Analysis: A 1:200 dilution from a representative lot detected NF-E2 related factor 2 in cells untreated and treated with hydrogen peroxide (Note: Cytoplasmic staining is increased under oxidative stress.).

Quality

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected NF-E2 related factor 2 in 10 µg of Ramos cell lysate.

Target description

~75 kDa observed. Two isoforms at 68 kDa (isoform 1) and 66 kDa (isoform 2) are known to exist. This protein has been observed at ~75 kDa due to the abundance of acidic residues found in Nrf2, which causes it to migrate anomalously in SDS-PAGE (Abu-Bakar, A., et al. (2007) Drug Metab Dispos. 35(5):787-794.). A ubiquitinated form of NF-E2-related factor 2 may be observed at ~110 kDa (Li, J., et al. (2005). Toxicol Sci. 83(2):313-328.).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Ramos cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Addition of Spirulina to Craft Beer: Evaluation of the Effects on Volatile Flavor Profile and Cytoprotective Properties.
Taiti, et al.
Antioxidants (Basel, Switzerland), 12 (2023)
Elena Frigato et al.
Oxidative medicine and cellular longevity, 2020, 2309437-2309437 (2020-05-08)
Circadian rhythms are biological oscillations that occur with an approximately 24 h period and optimize cellular homeostasis and responses to environmental stimuli. A growing collection of data suggests that chronic circadian disruption caused by novel lifestyle risk factors such as shift
Yuxia Li et al.
Molecules and cells, 37(3), 213-219 (2014-03-13)
MicroRNAs (miRNAs) represent a class of small non-coding regulatory RNAs that play important roles in normal hematopoiesis, including erythropoiesis. Although studies have identified several miRNAs that regulate erythroid commitment and differentiation, we do not understand the mechanism by which the
Sandra Gegunde et al.
ACS chemical neuroscience, 12(13), 2336-2346 (2021-06-11)
Anhydroexfoliamycin, a secondary metabolite from Streptomyces, has shown antioxidant properties in primary cortical neurons reducing neurodegenerative hallmarks diseases, both in vitro and in vivo models. Activated microglia, in the central nervous system, plays a crucial role in neuroinflammation and is
Nami Nakagomi et al.
Brain pathology (Zurich, Switzerland), 30(6), 1119-1133 (2020-07-21)
Epithelioid glioblastoma (E-GBM) was recently designated as a subtype of glioblastoma (GBM) by the World Health Organization (2016). E-GBM is an aggressive and rare variant of GBM that primarily occurs in children and young adults. Although most characterized cases of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service