Skip to Content
Merck
All Photos(2)

Key Documents

ABE203

Sigma-Aldrich

Anti-JARID1D Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysine-specific demethylase 5D, Histocompatibility Y antigen, H-Y, Histone demethylase JARID1D, Jumonji/ARID domain-containing protein 1D, Protein SmcY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5D(8284)

General description

Also known as Lysine-specific demethylase 5D, this histone demethylase is specific for ′Lys-4′ of histone H3 and is known to play a central role in histone function and spermatogenesis. It demethylates trimethylated and dimethylated, but not monomethylated H3 ′Lys-4′. Uniprot.org states two different isoforms of this enzyme.

Immunogen

KLH-conjugated linear peptide corresponding to human JARID1D.

Application

Immunocytochemistry Analysis: 2 µg/mL from a representative lot detected JARID1D in mESCs.

Immunoprecipitation Analysis: A representative lot immunoprecipitated JARID1D in human testis tissue lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-JARID1D Antibody (rabbit polyclonal antibody) validated in WB, ICC, IP to detect JARID1D also known as Lysine-specific demethylase 5D, Histocompatibility Y antigen, H-Y, Histone demethylase JARID1D.

Quality

Evaluated by Western Blot in human testis tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected JARID1D in 10 µg of human testis tissue lysate.

Target description

~174 kDa observed. Two isoforms may be observed due to alternative splicing at a molecular weight of ~167 kDa and ~174 kDa in some cell lysates. An uncharacterized band at a molecular weight of ~255 kDa may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human testis tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn Gunn et al.
Cancer discovery, 13(6), 1478-1497 (2023-02-28)
Oncogenic mutations in isocitrate dehydrogenase 1 (IDH1) and IDH2 occur in a wide range of cancers, including acute myeloid leukemia (AML) and glioma. Mutant IDH enzymes convert 2-oxoglutarate (2OG) to (R)-2-hydroxyglutarate [(R)-2HG], an oncometabolite that is hypothesized to promote cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service