ABE1076
Anti-Misu/NSun2 Antibody
serum, from rabbit
Synonym(s):
tRNA (cytosine(34)-C(5))-methyltransferase, Myc-induced SUN domain-containing protein, Misu, NOL1/NOP2/Sun domain family member 2, Substrate of AIM1/Aurora kinase B, tRNA (cytosine-5-)-methyltransferase, tRNA methyltransferase 4 homolog, hTrm4
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... NSUN2(54888)
Related Categories
General description
Immunogen
Application
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Misu/NSun2 in human epidermis tissue (Fry, M., and Watt, F. M. (2006). Curr Biol. 16(10):971-981.).
Epigenetics & Nuclear Function
RNA Metabolism & Binding Proteins
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Misu/NSun2 in 10 µg of HeLa cell lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service