Skip to Content
Merck
All Photos(1)

Key Documents

AB907

Sigma-Aldrich

Anti-Desmin Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, bovine, chicken, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DES(1674)

Specificity

Desmin. Reacts specifically with desmin in cultured cells or tissue preparations originating from chicken, human, bovine and mouse tissue. AB907 specifically stains the wide desmin band in immunoblot at a molecular weight of 50-55 kD.

Immunogen

Purified desmin from chicken gizzard, isolated by modified procedure of Geisler, et al [J. Biol Chem. (1980). 111:425-433].

Application

Anti-Desmin Antibody is an antibody against Desmin for use in WB, IH(P).
Indirect immunofluorescence of formalin fixed paraffin-embedded sections of human intestine sections: 1:10-1:20.

Immunoblotting

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Linkage

Replaces: 04-585

Physical form

Rabbit antiserum. Liquid containing 0.1% sodium azide as a preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yee Kit Tai et al.
Cells, 13(5) (2024-03-13)
Briefly (10 min) exposing C2C12 myotubes to low amplitude (1.5 mT) pulsed electromagnetic fields (PEMFs) generated a conditioned media (pCM) that was capable of mitigating breast cancer cell growth, migration, and invasiveness in vitro, whereas the conditioned media harvested from
Federica Maione et al.
Methods in molecular biology (Clifton, N.J.), 1267, 349-365 (2015-02-01)
Angiogenesis, the formation of neo-vessels, is one of the most important hallmarks of cancer. Tumor vasculature presents structural abnormalities such as dilatation of vessel diameter and hyper-branched and twisted pattern. A promising strategy in anticancer therapy to overcome the resistance
Witold W Kilarski et al.
Laboratory investigation; a journal of technical methods and pathology, 85(5), 643-654 (2005-02-22)
Migration, proliferation and invasive growth of myofibroblasts are key cellular events during formation of granulation tissue in situations of wound healing, arteriosclerosis and tumor growth. To study the invasive phenotype of myofibroblasts, we established an assay where arterial tissue from
Richard L Benton et al.
The Journal of comparative neurology, 507(1), 1031-1052 (2007-12-20)
After traumatic spinal cord injury (SCI), disruption and plasticity of the microvasculature within injured spinal tissue contribute to the pathological cascades associated with the evolution of both primary and secondary injury. Conversely, preserved vascular function most likely results in tissue
G P Sui et al.
BJU international, 90(1), 118-129 (2002-06-26)
To determine whether suburothelial interstitial cells of the human bladder express gap junctions, and if so, to establish their extent and composition, using immunocytochemistry, confocal microscopy and electron microscopy. Bladder tissue was obtained at cystectomy; the tissue was: (i) frozen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service