Skip to Content
Merck
All Photos(3)

Documents

AB5733

Sigma-Aldrich

Anti-Vimentin Antibody

serum, Chemicon®

Synonym(s):

Anti-F5H288

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, chicken, feline, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

Specificity

Recognizes Vimentin.

Immunogen

Recombinant vimentin.

Application

Detect Vimentin using this Anti-Vimentin Antibody validated for use in IC.
Immunocytochemistry: 1:1,000-1:5,000

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Physical form

Chicken IgY in PBS with 5 mM sodium azide as a preservative.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Breast carcinoma tissue, Colon, Prostate, Tonsil, Tonsilar lymphoma, Uterus, Ureter

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mammary stem cell population identified and characterized in late embryogenesis reveals similarities to human breast cancer.
Spike, BT; Engle, DD; Lin, JC; Cheung, SK; La, J; Wahl, GM
Cell Stem Cell null
GFAP isoforms in adult mouse brain with a focus on neurogenic astrocytes and reactive astrogliosis in mouse models of Alzheimer disease.
Kamphuis, W; Mamber, C; Moeton, M; Kooijman, L; Sluijs, JA; Jansen, AH; Verveer et al.
Testing null
Endotoxin-induced inflammation down-regulates L-type amino acid transporter 1 (LAT1) expression at the blood-brain barrier of male rats and mice.
Wittmann, G; Mohacsik, P; Balkhi, MY; Gereben, B; Lechan, RM
Fluids and Barriers of the Cns null
Martina Moeton et al.
Cellular and molecular life sciences : CMLS, 73(21), 4101-4120 (2016-05-05)
Glial fibrillary acidic protein (GFAP) is the characteristic intermediate filament (IF) protein in astrocytes. Expression of its main isoforms, GFAPα and GFAPδ, varies in astrocytes and astrocytoma implying a potential regulatory role in astrocyte physiology and pathology. An IF-network is
Irene Cimino et al.
Nature communications, 7, 10055-10055 (2016-01-13)
Anti-Müllerian hormone (AMH) plays crucial roles in sexual differentiation and gonadal functions. However, the possible extragonadal effects of AMH on the hypothalamic-pituitary-gonadal axis remain unexplored. Here we demonstrate that a significant subset of GnRH neurons both in mice and humans

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service