Skip to Content
Merck
All Photos(1)

Documents

AB5062P

Sigma-Aldrich

Anti-Vesicular GABA Transporter (VGAT) Antibody

Chemicon®, from rabbit

Synonym(s):

Vesicular Inhibitory Amino Acid Transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, monkey, human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Vesicular GABA Transporter (unc-47) homologue is a 525 amino acid long protein belonging to the family of neurotransmitter transporter (1). VGAT functions in the uptake of GABA and glycine into synaptic vesicles. Since the protein is bound to synaptic vesicles, VGAT antibodies are excellent markers of presynaptic GABAergic and glycinergic neurons.

Specificity

Antibody cross-reactivity with various species is not known. The immunogen sequence is 100% conserved in mouse, human and monkey.
Specific for Vesicular GABA Transporter (VGAT). Vesicular GABA Transporter (unc-47) homologue is a 525 amino acid long protein belonging to the family of neurotransmitter transporter. The immunogen peptide shows no significant homology with other known proteins.

Immunogen

A 17 amino acid peptide sequence near the carboxy terminal region of rat VGAT (McIntire, S., 1997).

Application

Immunohistochemistry/Immunofluorescence:
2-20 µg/mL of a previous lot was used on paraformaldehyde fixed sections.

ELISA:
1:10,000 – 1:100,000 dilution of a previous lot was used on ELISA, using 50-100 ng of control peptide (Cat. No. AG379) per well.

Western blot:
1-10 µg/mL (Chemiluminescence technique) The expected molecular weight is approximately 55-60 kDa.

Optimal working dilutions must be determined by the end user.
This Anti-Vesicular GABA Transporter (VGAT) Antibody is validated for use in ELISA, IF, IH, WB for the detection of Vesicular GABA Transporter (VGAT).

Quality

Evaluated by Western Blot on Rat retina lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected Vesicular GABA Transporter on 10μg of rat retina lysates.

Target description

57 kDa

Physical form

Purified rabbit polyclonal in buffer containing PBS with 0.1% BSA.

Analysis Note

Control
Brain tissue, rat retina lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phillip R Kramer et al.
Neuroscience, 356, 207-216 (2017-05-28)
Pain is a common complication of herpes zoster (HZ) infection which results from reactivation of a latent varicella zoster virus (VZV). A third of HZ patients' progress to a chronic pain state known as post herpetic neuralgia (PHN), and about
Ricardo Gómez-Nieto et al.
The Journal of comparative neurology, 516(4), 241-263 (2009-07-28)
Geometry of the dendritic tree and synaptic organization of afferent inputs are essential factors in determining how synaptic input is integrated by neurons. This information remains elusive for one of the first brainstem neurons involved in processing of the primary
Pablo Henny et al.
The Journal of comparative neurology, 499(4), 645-661 (2006-10-10)
Orexin/hypocretin (Orx) neurons are critical for the maintenance of waking in association with behavioral arousal and postural muscle tone, since with their loss narcolepsy with cataplexy occurs. Given that basal forebrain (BF) neurons project to the hypothalamus and play important
Zhihui Liu et al.
Neuron, 95(1), 106-122 (2017-07-07)
During social transmission of food preference (STFP), mice form long-term memory of food odors presented by a social partner. How does the brain associate a social context with odor signals to promote memory encoding? Here we show that odor exposure
Pablo Henny et al.
The European journal of neuroscience, 27(3), 654-670 (2008-02-19)
The present study was undertaken to characterize the pre- and postsynaptic constituents of the basal forebrain (BF) projection to the prefrontal cortex in the rat, and determine whether it includes glutamatergic in addition to established gamma-aminobutyric acid (GABA)ergic and cholinergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service