Skip to Content
Merck
All Photos(1)

Key Documents

AB4125

Sigma-Aldrich

Anti-FOXA2 Antibody

Chemicon®, from rabbit

Synonym(s):

Forkhead Box Protein A2, Hepatocyte Nuclear Factor 3-beta, HNF3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXA2(3170)
mouse ... Foxa2(15376)

Specificity

Recognizes FOXA2. The calculated molecular weight is ~48.3 kDa.

Immunogen

Synthetic peptide corresponding to amino acids 350-361 of human and rat FOXA2 (LGPPHHPGLPPE).

Application

Anti-FOXA2 Antibody is a Rabbit Polyclonal Antibody for detection of FOXA2 also known as Forkhead Box Protein A2, Hepatocyte Nuclear Factor 3-beta, HNF3B & has been validated in ICC.
Immunocytochemistry: 1/200 to 1/1000.

Optimal dilutions must be determined by the user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Affinity purified immunogloublin precipitated in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Maintain unopened vial at -20°C for up to 6 months. Avoid repeated freeze/thaw cycles.

The rehydrated antibody solutions can be stored undiluted at 2-8°C for 2 months without any significant loss of activity. Note, the solution is not sterile, thus care should be taken if product is stored at 2-8°C.

For storage at -20°C, the addition of an equal volume of glycerol can be used, however, it is recommended that ACS grade or higher glycerol be used, as significant loss of activity can occur if the glycerol used is not of high quality.

For freezing, it is recommended that the rehydrated antibody solution be further diluted 1:1 with a 2% BSA (fraction V, highest-grade available) solution made with the rehydration buffer. The resulting 1% BSA/antibody solution can be aliquoted and stored frozen at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles

PREPARATION AND USE:

To reconstitute the antibody, centrifuge the antibody vial at moderate speed (5,000 rpm) for 5 minutes to pellet the precipitated antibody product. Carefully remove the ammonium sulfate/PBS buffer solution and discard. It is not necessary to remove all of the ammonium sulfate/PBS solution: 10 μL of residual ammonium sulfate solution will not effect the resuspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur.

Resuspend the antibody pellet in any suitable biological buffer, standard PBS or TBS (pH 7.3-7.5) are typical. Volumes required are not critical but it is suggested that the final antibody concentration be between 0.1 mg/mL and 1.0 mg/mL. For example, to achieve a1 mg/mL concentration with 50 μg of precipitated antibody, the amount of buffer needed would be 50 μL.

Carefully add the liquid buffer to the pellet. DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to resuspend are normal. Vials are overfilled to compensate for any losses.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdulshakour Mohammadnia et al.
Journal of cellular physiology, 231(9), 1994-2006 (2016-01-13)
The generation of definitive endoderm (DE) from pluripotent stem cells (PSCs) is a fundamental stage in the formation of highly organized visceral organs, such as the liver and pancreas. Currently, there is a need for a comprehensive study that illustrates
Maryam Mahmoodinia Maymand et al.
Artificial cells, nanomedicine, and biotechnology, 46(4), 853-860 (2017-07-12)
The application of stem cells holds great promises in cell and tissue transplants. This study was designed to compare the hepatogenic differentiation of iPSCs on aligned PES/COL versus random. Aligned and random PES/COL nanofibrus scaffolds were fabricated by electrospining and
Saiyong Zhu et al.
Nature protocols, 10(7), 959-973 (2015-06-05)
Induction of tissue-specific cell types via a conventional transdifferentiation strategy typically uses overexpression of the corresponding lineage-specific transcription factors. Alternatively, somatic cells can be temporarily activated via a common set of reprogramming factors into a transition state, which can then
Afshin Noori et al.
Cell journal, 25(6), 372-382 (2023-07-12)
Efficient production of functional and mature alveolar epithelial is a major challenge for developing any cell replacement therapy for lung degenerative diseases. The extracellular matrix (ECM) pro-vides a dynamic environment and mediates cellular responses during development and maintenance of tissue
Ninuo Xia et al.
Scientific reports, 6, 20270-20270 (2016-02-05)
Generation of induced dopaminergic (iDA) neurons may provide a significant step forward towards cell replacement therapy for Parkinson's disease (PD). To study and compare transcriptional programs of induced cells versus primary DA neurons is a preliminary step towards characterizing human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service