Skip to Content
Merck
All Photos(1)

Key Documents

AB3540P

Sigma-Aldrich

Anti-Monocarboxylate Transporter 1 Antibody

Chemicon®, from rabbit

Synonym(s):

MCT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A1(6566)

Specificity

Recognizes rat Monocarboxylate Transporter 1 (MCT1). The immunogen shows no significant sequence homology with MCT2-8.

SPECIES REACTIVITIES: The immunogen sequence is 93% conserved in hamster, 80% in mouse and only 40% in human. Reactivity with other species has not been confirmed.

Immunogen

A 17 amino acid peptide sequence near the C-terminus of rat MCT1 (Gerhart et al. 1997; Knott et al. 1999; Takanaga et al. 1995; Carpenter et al. 1996; Koehler-Stec et al. 1998; Yoon et al. 1999; Halestrap & Price 1999).

Application

Detect Monocarboxylate Transporter 1 using this Anti-Monocarboxylate Transporter 1 Antibody validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-5 μg/mL using ECL. Membrane preps are recommended.



ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in PBS containing 0.1% BSA .

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changzheng Gao et al.
International journal of molecular medicine, 38(6), 1823-1830 (2016-10-26)
The lactate signaling cascade has recently been linked to mitochondrial energy metabolism and cardiomyocyte apoptosis in several cardiovascular diseases. Asiatic acid (AA) exhibits a variety of pharmacological effects, including antioxidant and anti-apoptotic effects. In this study, we investigated the protective effects
Vivian Rodriguez-Cruz et al.
Biopharmaceutics & drug disposition, 42(8), 351-358 (2021-07-01)
Gamma hydroxybutyric acid (GHB) has been approved clinically to treat excessive daytime sleepiness and cataplexy in patients with narcolepsy, alcohol and opioid withdrawal, and as an anesthetic. The use of GHB clinically is limited due to its high abuse potential.
Laurent A Messonnier et al.
Nutrients, 14(3) (2022-03-13)
It remains unclear whether sickle cell trait (SCT) should be considered a risk factor during intense physical activity. By triggering the polymerization-sickling-vaso-occlusion cascade, lactate accumulation-associated acidosis in response to high-intensity exercise is believed to be one of the causes of
Yan Sun et al.
Theranostics, 10(18), 8430-8445 (2020-07-30)
A viewpoint considering Alzheimer's disease (AD) as "type 3 diabetes" emphasizes the pivotal role of dysfunctional brain energy metabolism in AD. The hormone fibroblast growth factor 21 (FGF21) is a crucial regulator in energy metabolism; however, our understanding of the
Abdulrahman Alhamyani et al.
Journal of endocrinology and diabetes, 8(1) (2020-01-01)
The ventromedial hypothalamic nucleus (VMN) glucoregulatory neurotransmitters γ-aminobutyric acid (GABA) and nitric oxide (NO) signal adjustments in glycogen mobilization. Glucocorticoids control astrocyte glycogen metabolism in vitro. The classical (type II) glucocorticoid receptor (GR) is expressed in key brain structures that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service