Skip to Content
Merck
All Photos(1)

Key Documents

AB2231

Sigma-Aldrich

Anti-Dopamine Transporter; 100 µg Antibody

from rabbit

Synonym(s):

DA transporter, dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine),member 3, variable number tandem repeat (VNTR)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Plasmalemmal neurotransmitter transporters sequester synaptic and peri synaptic transmitter into presynaptic elements. The Dopamine Transporter (DAT), a 12 transmembrane domain transporter with the N and C terminal regions located within the cytoplasm is responsible for the reaccumulation of dopamine after it has been released. Levels of DAT protein expression are altered by chronic drug administration. The activity of the DAT reuptake carrier is sodium dependent, and it is suspected to play a role in such neurologic and psychiatric disorders as Parkinson′s disease, Tourette′s disease, schizophrenia, and addiction. DAT is also the target of several “DAT-blockers” including amphetamines and cocaine. These chemicals inhibit the action of DAT and, to a lesser extent, the other monoamine transporters, but their effects are mediated by separate mechanisms.

Specificity

Cat. AB2231 recognizes the N-terminus of the reduced and non reduced forms of DAT.

Application

This Anti-Dopamine Transporter, 100 µg Antibody is validated for use in WB for the detection of Dopamine Transporter, 100 µg.

Quality

Routinelyevaluated by western blot on rat brain lysate.

Target description

~ 50 – 80 kDa
Reduced form at approx 50 kDa and Non-reduced form at approx. 80 kDa

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Traumatic brain injury and trichloroethylene exposure interact and produce functional, histological, and mitochondrial deficits.
Sauerbeck, A; Hunter, R; Bing, G; Sullivan, PG
Experimental neurology null
Dafina M Angelova et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(2), 995-1006 (2017-10-22)
α-Synuclein (α-syn) is associated with a range of diseases, including Parkinson disease. In disease, α-syn is known to aggregate and has the potential to be neurotoxic. The association between copper and α-syn results in the formation of stellate toxic oligomers
Daniel W Curry et al.
Neuropharmacology, 128, 196-206 (2017-10-11)
S,R(+/-)-3,4-methylenedioxymethamphetamine (SR-MDMA) is an amphetamine derivative with prosocial and putative therapeutic effects. Ongoing clinical trials are investigating it as a treatment for post-traumatic stress disorder (PTSD) and other conditions. However, its potential for adverse effects such as hyperthermia and neurotoxicity
Gaia Faustini et al.
Neurobiology of disease, 138, 104789-104789 (2020-02-08)
Loss of dopaminergic nigrostriatal neurons and fibrillary α-synuclein (α-syn) aggregation in Lewy bodies (LB) characterize Parkinson's disease (PD). We recently found that Synapsin III (Syn III), a phosphoprotein regulating dopamine (DA) release with α-syn, is another key component of LB
Marion France et al.
Experimental physiology, 101(1), 81-99 (2015-09-19)
Obesity alters gastrointestinal (GI) motility and 5-HT signalling. Altered 5-HT signalling disrupts control of GI motility. Levels of extracellular 5-HT depend on enterochromaffin (EC) cell release and serotonin transporter (SERT) uptake. We assessed GI transit and 5-HT signalling in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service