AB17003
Anti-Bim Antibody, internal epitope, pan-Bim isoforms
Chemicon®, from rabbit
Synonym(s):
BOD
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human, rat
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... TMBIM4(51643)
Specificity
Recognizes pan-Bim isoforms.
Immunogen
Epitope: internal epitope, pan-Bim isoforms
Synthetic peptide corresponding to amino acids 22-40 of human origin (O′Conneor et al. 1998). The sequence is identical to that of mouse and is different by one amino acid from that of rat.
Application
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
BCL2 & Inhibition
This Anti-Bim Antibody, internal epitope, pan-Bim isoforms is validated for use in WB for the detection of Bim.
Western blot: 1:1000-1:2000
Human K562 cell lysate can be used as a positive control and a 23 kDa band can be detected.
Optimal working dilutions must be determined by end user.
Human K562 cell lysate can be used as a positive control and a 23 kDa band can be detected.
Optimal working dilutions must be determined by end user.
Target description
23 kDa
Physical form
Format: Purified
ImmunoAffinity Purified
Immunoaffinity Purified immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Storage and Stability
The undiluted antibody solution is stable for approximately 6 months when stored 2–8°C.
Analysis Note
Control
Skin/Skin Tumor
Skin/Skin Tumor
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
NF-Y is essential for expression of the proapoptotic bim gene in sympathetic neurons.
Hughes, R; Kristiansen, M; Lassot, I; Desagher, S; Mantovani, R; Ham, J
Cell Death and Differentiation null
Julia P Huang et al.
Oncology letters, 14(5), 5735-5742 (2017-11-09)
Enhancer of zeste homolog 2 (EZH2), a subunit of polycomb repressive complex 2, is a histone methyl-transferase and is considered to work cooperatively with histone deacetylases (HDACs) in the same protein complex to mediate gene transcription repression by increasing histone
A Malyukova et al.
Leukemia, 27(5), 1053-1062 (2012-12-12)
Loss of function mutation in FBXW7, an E3 ubiquitin ligase, is associated with good prognosis and early glucocorticoid treatment response in childhood T-cell acute lymphoblastic leukemia (T-ALL) by unknown mechanisms. Here, we show that FBXW7 targets the glucocorticoid receptor α
Shannon M Matulis et al.
Molecular cancer therapeutics, 8(5), 1197-1206 (2009-05-07)
Here, we report on the organic arsenical darinaparsin (ZIO-101, S-dimethylarsino-glutathione) and its anti-myeloma activity compared with inorganic arsenic trioxide. Darinaparsin induced apoptosis in multiple myeloma cell lines in a dose-dependent manner, and the addition of N-acetylcysteine, which increases intracellular glutathione
Min Ae Han et al.
Oncotarget, 7(40), 65707-65720 (2016-09-02)
Eupafolin, a flavone found in Artemisia princeps, has been reported for its anti-tumor activity in several cancer cells. In this study, we examined whether eupafolin could sensitize TRAIL-mediated apoptosis in human renal carcinoma Caki cells. We found that eupafolin alone
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service