Skip to Content
Merck
All Photos(3)

Documents

AB1031

Sigma-Aldrich

Anti-Aggrecan Antibody

Chemicon®, from rabbit

Synonym(s):

Cartilage-specific proteoglycan core protein, Cartilage-specific proteoglycan core protein, aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122), aggrecan proteoglycan, aggrecan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

General description

Aggregating chondroitin sulfate proteoglycan or aggrecan, is one of the most studied proteoglycans. Many individual monomers of aggrecan bind to hyaluronic acid to form an aggregate, the monomer is known as aggrecan. The aggregates are comprised of up to 100 monomers attached to a single chain of hyaluronic acid. The high content of negatively charged polysaccharide chains contributes to the osmotic swelling of cartilage and in combination with intact collagen fibers results in articular cartilage resistance to compressive forces and high tensile strength. Aggrecan is abundant in articular cartilage and compromises 10% of its dry weight.

Specificity

AB1031 is specific for mouse aggrecan; other species have not been tested.
Expected to cross react with Human and rat.

Immunogen

GST fusion protein containing amino acids 1177-1326 of mouse aggrecan.

Application

Anti-Aggrecan Antibody is an antibody against Aggrecan for use in IF, IH(P), IP & WB.
Immunohistochemistry(Paraffin):
A previous lot of this antibody was used at 10 μg/mL. See note under Protocol regarding triton X-100 treatments.

Immunoprecipitation:
A previous lot of this antibody was used at 5 μg/ 500 μL of cell membrane extract in PBS with inhibitors and >0.5% triton X-100.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Routinely evaluated by Western Blot on Mouse Brain lysates.

Western Blot Analysis:
1:500 dilution of this lot detected AGGRECAN on 10 μg of Mouse Brain lysates.

Physical form

ImmunoAffinity Purified
Rabbit polyclonal antigen affinity purified antibody in PBS. Anti-GST has not been adsorbed. Liquid.
Note: Aggrecan is present in articular cartilage, brain, heart, and other tissues.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Cartilage, neural tube, and brain tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Biglycan and fibromodulin have essential roles in regulating chondrogenesis and extracellular matrix turnover in temporomandibular joint osteoarthritis.
Embree, MC; Kilts, TM; Ono, M; Inkson, CA; Syed-Picard, F; Karsdal, MA; Oldberg, A; Bi, Y; Young, MF
The American Journal of Pathology null
Timothy P Rutkowski et al.
Journal of cell science, 129(11), 2145-2155 (2016-05-11)
RBPjκ-dependent Notch signaling regulates multiple processes during cartilage development, including chondrogenesis, chondrocyte hypertrophy and cartilage matrix catabolism. Select members of the HES- and HEY-families of transcription factors are recognized Notch signaling targets that mediate specific aspects of Notch function during
Mandy Sonntag et al.
BMC biology, 16(1), 99-99 (2018-09-27)
Perineuronal nets (PNNs) are specialized aggregations of extracellular matrix (ECM) molecules surrounding specific neurons in the central nervous system (CNS). PNNs are supposed to control synaptic transmission and are frequently associated with neurons firing at high rates, including principal neurons
Huimin Yan et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(41), E6199-E6208 (2016-09-30)
Osteoarthritis (OA) is a major cause of disability and morbidity in the aging population. Joint injury leads to cartilage damage, a known determinant for subsequent development of posttraumatic OA, which accounts for 12% of all OA. Understanding the early molecular
Luigi A Nasto et al.
Mechanisms of ageing and development, 134(1-2), 35-42 (2012-12-25)
Intervertebral disc degeneration (IDD) is the leading cause of debilitating spinal disorders such as chronic lower back pain. Aging is the greatest risk factor for IDD. Previously, we demonstrated IDD in a murine model of a progeroid syndrome caused by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service