Skip to Content
Merck
All Photos(2)

Key Documents

AAWG0250C

Millipore

MCE Membrane Filter, 0.8 μm Pore Size

MF-Millipore, filter diam. 25 mm, hydrophilic, white, gridded, 150 μg/cm2 binding capacity (protein)

Synonym(s):

MF-Millipore Membrane Filter, mixed cellulose esters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.22

material

gridded
mixed cellulose esters (MCE) membrane
white filter

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

25 mm

thickness

150 μm

gravimetric extractables

4%

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.8 μm pore size
82 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥1.0 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

  • MF-Millipore mixed cellulose esters membrane filter is ideal for biological and environmental monitoring applications.
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods.
  • The white filter is available with a gridded surface.
MF-Millipore membrane filters are made up of a biologically inert mix of cellulose acetate and cellulose nitrate. MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have lower water extractable content than standard MF-Millipore filters.
This 25 mm MF-Millipore membrane is composed of mixed cellulose membrane & has a 0.8 um pore size. With biologically inert mixtures of cellulose acetate & cellulose nitrate, MF-Millipore membranes are widely used in analytical/research applications.

Application

  • Air monitoring
  • Particle monitoring
  • Particle removal
  • Bioassays
  • Microplastics

Features and Benefits

  • Versatile filter
  • Available in a range of pore sizes
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service