8.52067
Fmoc-Arg(Pbf)-OH
Novabiochem®
Synonym(s):
Fmoc-Arg(Pbf)-OH, N-α-Fmoc-N G-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine
About This Item
Quality Level
product line
Novabiochem®
Assay
≥98% (TLC)
≥99.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
mp
>65 °C
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
-10 to -25°C
SMILES string
[S](=O)(=O)(N\C(=N\CCC[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O)\N)c1c(c2c(c(c1C)C)OC(C2)(C)C)C
InChI
1S/C34H40N4O7S/c1-19-20(2)30(21(3)26-17-34(4,5)45-29(19)26)46(42,43)38-32(35)36-16-10-15-28(31(39)40)37-33(41)44-18-27-24-13-8-6-11-22(24)23-12-7-9-14-25(23)27/h6-9,11-14,27-28H,10,15-18H2,1-5H3,(H,37,41)(H,39,40)(H3,35,36,38)/t28-/m0/s1
InChI key
HNICLNKVURBTKV-NDEPHWFRSA-N
General description
The standard derivative for the introduction of Arg in Fmoc SPPS [1,2]. The Pbf side-chain protecting group is removed with TFA approximately 1-2 times faster than Pmc.In the preparation of peptides containing both Arg and Trp, it is recommended that this derivative is used in conjunction with Fmoc-Trp(Boc)-OH (852050).
Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS
Literature references
[1] L. A. Carpino, et al. (1993) Tetrahedron Lett., 34, 7829.
[2] C. G. Fields, et al. (1993) Tetrahedron Lett., 34, 6661.
Application
- Syntheses of cyanophycin segments for investigations of cell-penetration: Describes the synthesis process and overall yields for steps from Fmoc-Arg(Pbf)-OH to Adp building blocks. (M Grogg, D Hilvert, D Seebach, 2019).
- Revisiting NO2 as Protecting Group of Arginine in Solid-Phase Peptide Synthesis: Compares the stability of Fmoc-Arg(Boc) 2 -OH with Fmoc-Arg(Pbf)-OH in solvents over time. (M Alhassan, A Kumar, J Lopez, F Albericio, 2020).
- Gene delivery of PAMAM dendrimer conjugated with the nuclear localization signal peptide originated from fibroblast growth factor 3: Discusses a conjugation reaction involving Fmoc-Arg(pbf)-OH in gene delivery systems. (J Lee, J Jung, YJ Kim, E Lee, JS Choi, 2014).
Linkage
Analysis Note
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Arg(Pbf)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Arg(Pbf)-Arg(Pbf)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Arg-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (HPLC): ≤ 0.2 %
Purity (TLC(011C)): ≥ 98 %
Purity (TLC(157B)): ≥ 98 %
Solubility (12,5 mmol in 25 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 1.0 %
Acetate (IC): ≤ 0.02 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service