Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.22330

Sigma-Aldrich

Methyl benzoate

for synthesis

Synonym(s):

Methyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-259-7
NACRES:
NA.22

Pricing and availability is not currently available.

vapor pressure

0.36 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

510 °C

potency

1177 mg/kg LD50, oral (Rat)
2170 mg/kg LD50, skin (Rabbit)

expl. lim.

8.6-20 % (v/v)

bp

199 °C/1013 hPa

mp

-12 °C

transition temp

flash point 82 °C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
524632524631524624
manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

100

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage condition

OK to freeze, protect from light, desiccated (hygroscopic)

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, desiccated (hygroscopic)

storage condition

OK to freeze

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.086 - 1.088
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service