Skip to Content
Merck
All Photos(1)

Key Documents

8.22050

Sigma-Aldrich

Sodium dodecyl sulfate

for synthesis

Synonym(s):

Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Dodecyl sodium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25NaO4S
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-788-1
NACRES:
NA.22
form:
powder
solubility:
>130 g/L
Pricing and availability is not currently available.

Quality Level

form

powder

potency

1427 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6-9 (20 °C, 10 g/L in H2O)

mp

204-207 °C

transition temp

flash point >150 °C

solubility

>130 g/L

density

1.1 g/cm3 at 20 °C

bulk density

490‑560 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[S](=O)(=O)([O-])OCCCCCCCCCCCC

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Sodium dodecyl sulfate (SDS) is a conventional anionic surfactant. An amphiphilic surfactant consists of polar hydrophilic head groups and non-polar hydrophobic tail groups. Surfactants are characterized by their ability to form micelles, i.e., their ability to self-aggregate above a critical micelle concentration (CMC). Due to its amphiphilic nature, the surfactant is used as a detergent in industries. They are useful for cleaning, wetting, dispersing, emulsifying, foaming, and antifoaming.

Application

Sodium dodecyl sulfate used as a surfactant:
  • In the synthesis of SiO2/polystyrene nanocomposite particles by miniemulsion polymerization.
  • In the preparation of solid lipid NPs (SLNs) and poly(lactide-co-glycolide) NPs containing OA-Fe3O4 as CANs (composite antistatic NPs).

Analysis Note

Assay (Epton titration): ≥ 95.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service