Skip to Content
Merck
All Photos(1)

Key Documents

8.20978

Sigma-Aldrich

Diphenyl ether

for synthesis

Synonym(s):

Diphenyl ether, Phenyl ether, Diphenyl oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352112
EC Index Number:
202-981-2
NACRES:
NA.22
grade:
for synthesis
bp:
259 °C/1013 hPa
vapor pressure:
0.05 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

0.05 hPa ( 20 °C)

Assay

≥98.0% (GC)

form

solid

autoignition temp.

610 °C

potency

2450 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-1.5 % (v/v)

dilution

(for synthesis)

bp

259 °C/1013 hPa

mp

24-27 °C

transition temp

flash point 115 °C

solubility

0.018 g/L

density

1.07 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(c2ccccc2)c1ccccc1

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 24 °C
Melting range (upper value): ≤ 28 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service