Skip to Content
Merck
All Photos(1)

Key Documents

8.20931

Sigma-Aldrich

1-Octanol

for synthesis

Synonym(s):

1-Octanol, n-Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-917-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
195 °C/1013 hPa
vapor pressure:
0.031 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

0.031 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

270 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8 % (v/v)

dilution

(for synthesis)

kinematic viscosity

5.6 cSt(40 °C)

bp

195 °C/1013 hPa

transition temp

flash point 86 °C

solubility

0.43 g/L (External MSDS)

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCC

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Application

1-Octanol can be used as a reactant to synthesize:
  • Octenes via dehydration.
  • Esters via esterification with carboxylic acids.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.824 - 0.826
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service