8.18712
Hexamethylenetetramine
for synthesis
Synonym(s):
Hexamethylenetetramine, Hexamethylenetetramine, Hexamine, Formin, Urotropin
About This Item
vapor pressure
<0.01 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (perchloric acid titration)
form
solid
autoignition temp.
390 °C
potency
9200 mg/kg LD50, oral (Rat)
pH
7-10 (20 °C, 100 g/L in H2O)
transition temp
flash point 250 °C
solubility
soluble 895 g/L
density
1.33 g/cm3 at 20 °C
bulk density
600 kg/m3
storage temp.
2-30°C
SMILES string
N21CN3CN(C2)CN(C1)C3
InChI
1S/C6H12N4/c1-7-2-9-4-8(1)5-10(3-7)6-9/h1-6H2
InChI key
VKYKSIONXSXAKP-UHFFFAOYSA-N
Related Categories
Application
- Electrochemical detection of microplastics: Hexamethylenetetramine is utilized in a novel sensor design using chitosan and magnesium oxide for the electrochemical detection of microplastics, enhancing environmental monitoring capabilities (Noumani et al., 2024).
- Novel methodologies in nanoparticle synthesis: Research details the use of hexamethylenetetramine in the preparation of monodisperse cerium oxide nanoparticles, demonstrating its utility in precise nanoparticle fabrication (Hastings et al., 2024).
- Development in mesoporous materials: A study explores hexamethylenetetramine′s use in the self-assembly of ordered mesoporous nanofibers, contributing to advancements in materials science and engineering (Peng et al., 2023).
Analysis Note
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 2 - Skin Sens. 1B
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 1
Flash Point(F)
482.0 °F - closed cup
Flash Point(C)
250 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service