Skip to Content
Merck
All Photos(1)

Key Documents

8.18112

Sigma-Aldrich

Sulfur trioxide pyridine complex

for synthesis

Synonym(s):

Sulfur trioxide pyridine complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N · O3S
CAS Number:
Molecular Weight:
159.16
MDL number:
UNSPSC Code:
12352303
EC Index Number:
247-683-3
NACRES:
NA.22
Assay:
≥97.0% (acidimetric)

Quality Level

Assay

≥97.0% (acidimetric)

autoignition temp.

>350 °C

potency

500-2000 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

mp

155-165 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)=O.n1ccccc1

InChI

1S/C5H5N.O3S/c1-2-4-6-5-3-1;1-4(2)3/h1-5H;

InChI key

UDYFLDICVHJSOY-UHFFFAOYSA-N

Application


  • Post-synthetic modulation of the charge distribution in a metal–organic framework for optimal binding of carbon dioxide and sulfur dioxide: This study discusses the modulation of pore environment to optimize the binding of sulfur dioxide in metal-organic frameworks. (L Li et al., 2019).

  • Sulfation of abies ethanol lignin by complexes of sulfur trioxide with 1, 4-dioxane and pyridine: Research on the sulfation of lignin using a complex of sulfur trioxide with pyridine, exploring its implications and efficiency. (YN Malyar et al., 2021).

  • Antioxidant and anticoagulant activities of mycelia polysaccharides from Catathelasma ventricosum after sulfated modification: This article explores the use of sulfur trioxide–pyridine complex for sulfating polysaccharides, enhancing their biological activities. (Y Liu et al., 2018).

  • Sulfation of citrus pectin by pyridine-sulfurtrioxide complex and its anticoagulant activity: Study on derivatizing citrus pectin using a pyridine-sulfur trioxide complex to explore its potential health benefits. (Y Hu et al., 2015).

  • Sulfur Dioxide–Pyridine Dimer. FTIR and Theoretical Evidence for a Low-Symmetry Structure: An investigation into the structure of the sulfur dioxide–pyridine complex using FTIR spectroscopy and theoretical models. (JW Keller, 2015).

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 160 °C
Melting range (upper value): ≤ 170 °C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service