8.14483
Amidosulfuric acid
for synthesis
Synonym(s):
Amidosulfuric acid, Amidosulfonic acid, Sulphamic acid, Sulfamidic acid
About This Item
grade
for synthesis
synthesis grade
Quality Level
Assay
≥99% (acidimetric)
form
solid
pH
1.2 (10 g/L in H2O)
mp
205 °C (decomposition)
solubility
213 g/L
density
2.13 g/cm3 at 20 °C
bulk density
600 kg/m3
storage temp.
2-30°C
SMILES string
[S](=O)(=O)([N+H3])[O-]
InChI
1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)
InChI key
IIACRCGMVDHOTQ-UHFFFAOYSA-N
Analysis Note
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service