8.08326
4-Toluenesulfonyl chloride
for synthesis
Synonym(s):
4-Toluenesulfonyl chloride
About This Item
Quality Level
Assay
≥98.0% (GC)
form
solid
potency
4680 mg/kg LD50, oral (Rat)
mp
67 °C
transition temp
flash point 128 °C
density
1.49 g/cm3 at 20 °C
bulk density
800 kg/m3
storage temp.
2-30°C
SMILES string
[S](=O)(=O)(Cl)c1ccc(cc1)C
InChI
1S/C7H7ClO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3
InChI key
YYROPELSRYBVMQ-UHFFFAOYSA-N
Application
- Neurophysiological research on neurotransmitter receptors: Employing Dopamine-D4 hydrochloride solution in studies to investigate the postsynaptic inhibition of GABAergic currents by dopamine D4 receptors in the globus pallidus of mice (Shin et al., 2003).
Analysis Note
Melting range (lower value): ≥ 66 °C
Melting range (upper value): ≤ 70 °C
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
262.4 °F
Flash Point(C)
128 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service