Skip to Content
Merck
All Photos(1)

Key Documents

8.06838

Sigma-Aldrich

n-Nonane

for synthesis

Synonym(s):

n-Nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH3
CAS Number:
Molecular Weight:
128.26
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-913-4
NACRES:
NA.22
grade:
for synthesis
bp:
151 °C/1013 hPa
vapor pressure:
5 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor pressure

5 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

205 °C

expl. lim.

0.7-5.6 % (v/v)

dilution

(for synthesis)

kinematic viscosity

<7 cSt(40 °C)

bp

151 °C/1013 hPa

mp

-53 °C

transition temp

flash point 31 °C

density

0.72 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCC)CCCC

InChI

1S/C9H20/c1-3-5-7-9-8-6-4-2/h3-9H2,1-2H3

InChI key

BKIMMITUMNQMOS-UHFFFAOYSA-N

Application


  • Probing the nucleation mechanism for the binary n-nonane/1-alcohol series with atomistic simulations.: This study explores the nucleation mechanisms of n-Nonane in combination with 1-alcohols through detailed atomistic simulations. It provides insights into molecular interactions and phase behaviors, which are critical for applications in chemical synthesis and materials science (Nellas et al., 2006).

  • Enhancement of propranolol hydrochloride and diazepam skin absorption in vitro. II: Drug, vehicle, and enhancer penetration kinetics.: This research investigates the use of n-Nonane as a vehicle in enhancing the skin absorption of pharmaceutical compounds. It examines the kinetics of drug penetration and the effectiveness of n-Nonane in facilitating drug delivery through the skin (Hori et al., 1992).


Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 0.717 - 0.719
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service