Skip to Content
Merck
All Photos(1)

Key Documents

8.04608

Sigma-Aldrich

Hydrazinium hydroxide

(about 100% N₂H₅OH) for synthesis

Synonym(s):

Hydrazinium hydroxide, Hydrazine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4N2 · xH2O
CAS Number:
Molecular Weight:
32.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352305
EC Index Number:
206-114-9
NACRES:
NA.22
form:
liquid

vapor pressure

20 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

280 °C

potency

169 mg/kg LD50, oral (Rat)

expl. lim.

3.4-99 % (v/v)

pH

10-11 (20 °C, 10 g/L in H2O)

bp

120.5 °C/1013 hPa

mp

-51.7 °C

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[N+H3]N.[O-H]

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

Application

Hydrazinium hydroxide (N2H4.H2O) can be used as a reducing agent to synthesize:
  • Magnetic nickel nanoparticles by polyol process using nickel acetate as a precursor.
  • Cu@Cu2O core-shell nanoparticles via a chemical reduction method using CuSO4.5H2O as a precursor.

It can also be used as a reactant to prepare:
2H-indazolo[2,1-b]phthalazine-1,6,11(13H)-trione derivatives by Ce(SO4)2·4H2O catalyzed condensation reaction with phthalic anhydride, aromatic aldehydes, and dimedone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup - (anhydrous substance)

Flash Point(C)

38 °C - closed cup - (anhydrous substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service