Skip to Content
Merck
All Photos(1)

Key Documents

8.01816

Sigma-Aldrich

Benzyltrimethylammonium hydroxide

(40% solution in methanol) for synthesis

Synonym(s):

Trimethylbenzylammonium hydroxide, Triton B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

127 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

455 °C

expl. lim.

5.1-44 % (v/v)

pH

12 (20 °C in H2O, undiluted)

bp

65 °C/1013 hPa

mp

-98 °C

transition temp

flash point 15 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 39.0 - 43.0 %
Water (K. F.): ≤ 3.0 %
Colour number (Apha): ≤ 200
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

59.0 °F

Flash Point(C)

15 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service