Skip to Content
Merck
All Photos(1)

Key Documents

8.00470

Sigma-Aldrich

Benzenesulfonyl chloride

for synthesis

Synonym(s):

Benzenesulfonyl chloride, Benzene sulfochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClO2S
CAS Number:
Molecular Weight:
176.62
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-636-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

6 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

450 °C

potency

1615 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

252 °C/1013 hPa (decomposes)

mp

15 °C

transition temp

flash point 123 °C

density

1.38 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(Cl)c1ccccc1

InChI

1S/C6H5ClO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H

InChI key

CSKNSYBAZOQPLR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.377 - 1.379
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service