Skip to Content
Merck
All Photos(1)

Key Documents

71137

Sigma-Aldrich

Rosetta-gami B(DE3)pLysS Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

Rosetta-gami Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

Product Name

Rosetta-gami B(DE3)pLysS Competent Cells - Novagen, Rosetta-gami B strains combines the key features of BL21, Origami, and Rosetta to enhances the expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: 1.0×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: FompT hsdSB (rB mB) gal dcm lacY1 ahpC (DE3) gor522::Tn10 trxB pLysSRARE (CamR, KanR, TetR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Rosetta-gami B strains combine the key features of BL21 (and its Tuner derivative), Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds in the bacterial cytoplasm. These strains are compatible with ampicillin- or spectinomycin-resistant vectors.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Rosetta-gami B strains combines the key features of BL21, Origami, and Rosetta to enhances the expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosetta-gami B(DE3)pLysS Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•10 µl10 µlTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kalinka Koteva et al.
Cell chemical biology, 25(4), 403-412 (2018-02-06)
Rifamycin monooxygenases (Rox) are present in a variety of environmental bacteria and are associated with decomposition of the clinically utilized antibiotic rifampin. Here we report the structure and function of a drug-inducible rox gene from Streptomyces venezuelae, which encodes a
Christophe Macri et al.
Autophagy, 11(3), 472-486 (2015-02-27)
The P140 peptide, a 21-mer linear peptide (sequence 131-151) generated from the spliceosomal SNRNP70/U1-70K protein, contains a phosphoserine residue at position 140. It significantly ameliorates clinical manifestations in autoimmune patients with systemic lupus erythematosus and enhances survival in MRL/lpr lupus-prone
Keisuke Goto et al.
Diagnostic pathology, 17(1), 16-16 (2022-02-01)
The human amphoterin-induced gene and open reading frame (AMIGO) was identified as a novel cell adhesion molecule of type I transmembrane protein. AMIGO2 is one of three members of the AMIGO family (AMIGO1, 2, and 3), and the similarity between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service