Skip to Content
Merck
All Photos(1)

Key Documents

567540

Sigma-Aldrich

Sodium Orthovanadate

A broad spectrum potent inhibitor of protein tyrosine phosphatases.

Synonym(s):

Sodium Orthovanadate, PTP Inhibitor X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na3O4V
CAS Number:
Molecular Weight:
183.91
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

description

RTECS - YW1120000

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[V](=O)([O-])([O-])[O-].[Na+].[Na+].[Na+]

InChI

1S/3Na.4O.V/q3*+1;;3*-1;

InChI key

IHIXIJGXTJIKRB-UHFFFAOYSA-N

General description

A broad spectrum potent inhibitor of protein tyrosine phosphatases. Also known to inhibit Na+/K+ ATPase, acid and alkaline phosphatases, phosphofructokinase, and adenylate kinase. Most recently shown to inhibit ATPase activity of the reconstituted binding protein-dependent ATP-Binding Cassette (ABC) transporter for maltose (MalFGK2) of Salmonella typhimurium in the micromolar range. Can be converted to pervanadate by hydrogen peroxide.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Na+,K+-ATPase, acid , alkaline phosphatases, phosphofructokinase, and adenylate kinase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Huyer, G., et al. 1997. J. Biol. Chem. 272, 843.
Hunke, S., et al. 1995. Biochem. Biophys. Res. Commun.216, 589.
Levchuck, S.G., et al. 1994. Pediatric Res.37, 382A.
Fohr, K.J., et al. 1989. Biochem. J.262, 83.
Swarup, G., et al. 1982. Biochem. Biophys. Res. Commun.107, 1104.
Seargeant, L.E. and Stinson, R.A. 1979. Biochem. J.181, 247.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marwa Ben-Khemis et al.
Redox biology, 67, 102898-102898 (2023-09-28)
TNFα-mediated signaling pathways play a pivotal role in the pathogenesis of inflammatory diseases such as rheumatoid arthritis (RA) and inflammatory bowel disease (IBD) by promoting phagocyte inflammatory functions, notably cytokine release and reactive oxygen species (ROS) production by NOX2. In
Seungkirl Ahn et al.
The Journal of biological chemistry, 277(29), 26642-26651 (2002-05-16)
Endocytosis of ligand-activated receptors requires dynamin-mediated GTP hydrolysis, which is regulated by dynamin self-assembly. Here, we demonstrate that phosphorylation of dynamin I by c-Src induces its self-assembly and increases its GTPase activity. Electron microscopic analyses reveal that tyrosine-phosphorylated dynamin I
Louis Marois et al.
Journal of immunology (Baltimore, Md. : 1950), 182(4), 2374-2384 (2009-02-10)
Little is known about the mechanisms that arrest FcgammaRIIa signaling in human neutrophils once engaged by immune complexes or opsonized pathogens. In our previous studies, we observed a loss of immunoreactivity of Abs directed against FcgammaRIIa following its cross-linking. In
Silvia Peppicelli et al.
International journal of molecular sciences, 23(23) (2022-12-12)
Clinical outcomes of melanoma patients pointed out a gender disparity that supports a correlation between sex hormone activity on estrogen receptors (ER) and melanoma development and progression. Here, we found that the epithelial-to-mesenchymal transition (EMT) of melanoma cells induced by
Da-Yun Seo et al.
International journal of molecular sciences, 25(13) (2024-07-13)
Isoscopoletin is a compound derived from various plants traditionally used for the treatment of skin diseases. However, there have been no reported therapeutic effects of isoscopoletin on atopic dermatitis (AD). AD is a chronic inflammatory skin disease, and commonly used

Articles

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service