Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
5.43828
ortho-Phosphoric acid
85%, for HPLC LiChropur™
Synonym(s):
Phosphoric acid, o-Phosphoric acid, Orthophosphoric acid, Orthophosphoric acid
About This Item
Recommended Products
Product Name
ortho-Phosphoric acid 85%, for HPLC LiChropur™
vapor density
3.4 (vs air)
Quality Level
vapor pressure
2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)
form
liquid
technique(s)
HPLC: suitable
pH
<0.5 (20 °C, 100 g/L in H2O)
kinematic viscosity
30.5 cSt(20 °C)
bp
158 °C (lit.)
mp
~40 °C (lit.)
density
1.685 g/mL at 25 °C (lit.)
storage temp.
no temp limit
SMILES string
OP(O)(O)=O
InChI
1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)
InChI key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.
The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.
The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.
The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.
Protocols
Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.
Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.
Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.
Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service