Skip to Content
Merck
All Photos(1)

Key Documents

5.32996

Sigma-Aldrich

Androgen Receptor DBD Inhibitor, VPC-14449

Synonym(s):

Androgen Receptor DBD Inhibitor, VPC-14449, AR-DBD Inhibitor, VPC-14449, VPC14449, VPC 14449

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Br2N4OS
CAS Number:
Molecular Weight:
394.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

947 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

C1COCCN1C2=NC(=CS2)N3C=C(N=C3Br)Br

InChI

1S/C10H10Br2N4OS/c11-7-5-16(9(12)13-7)8-6-18-10(14-8)15-1-3-17-4-2-15/h5-6H,1-4H2

InChI key

AZLNRGRZOLVWRX-UHFFFAOYSA-N

General description

A cell-permeable morpholinyl thiazole compound that selectively binds to the DNA-binding domain of androgen receptor (AR) and blocks its activity (IC50 = 340 nM in PC3 cells). Its inhibitory effect is comparable to enzalutamide (IC50 = 314 nM). Does not affect Y594D and Q592D mutant forms of AR. However, Y594A and Q592A mutant AR activity can be inhibited at much higher concentrations (3 - 6 mM). Shown to down-regulate the expression of AR-responsive genes in LNCaP Cells, however, it does not affect genotoxin-responsive genes expression. Suppresses the growth of androgen-sensitive LNCaP xenografts in mice (~100 mg/kg, i.p., b.i.d.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable morpholinyl thiazole compound that selectively binds to the DNA-binding domain of androgen receptor (AR) and blocks its activity (IC50 = 340 nM in PC3 cells). Its inhibitory effect is comparable to enzalutamide (IC50 = 314 nM). Does not affect Y594D and Q592D mutant forms of AR. However, Y594A and Q592A mutant AR activity can be inhibited at much higher concentrations (3 - 6 mM). Shown to down-regulate the expression of AR-responsive genes in LNCaP Cells, however, it does not affect genotoxin-responsive genes expression. Suppresses the growth of androgen-sensitive LNCaP xenografts in mice (~100 mg/kg, i.p., b.i.d.).



Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
VPC-14449, is a cell-permeable compound that selectively binds to the DNA-binding domain of androgen receptor (AR) and blocks its activity (IC₅₀ = 340 nM in PC3 cells).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Androgen Receptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dalal, K., et al. 2014. J. Biol. Chem.289, 26417.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service