Skip to Content
Merck
All Photos(2)

Key Documents

5.31167

Sigma-Aldrich

GSK-3 Inhibitor XXIX, CHIR98014

Synonym(s):

GSK-3 Inhibitor XXIX, CHIR98014, N²-(2-(4-(2,4-Dichlorophenyl)-5-(1H-imidazol-1-yl)pyrimidin-2-ylamino)ethyl)-5-nitropyridine-2,6-diamine, N⁶-(2-((4-(2,4-Dichlorophenyl)-5-(1H-imidazol-1-yl)-2-pyrimidinyl)amino}ethyl)-3-nitro-2,6-pyridinediamine, CHIR-98014, CT-98014

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17Cl2N9O2
CAS Number:
Molecular Weight:
486.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

InChI

1S/C20H17Cl2N9O2/c21-12-1-2-13(14(22)9-12)18-16(30-8-7-24-11-30)10-27-20(29-18)26-6-5-25-17-4-3-15(31(32)33)19(23)28-17/h1-4,7-11H,5-6H2,(H3,23,25,28)(H,26,27,29)

InChI key

MDZCSIDIPDZWKL-UHFFFAOYSA-N

General description

A cell-permeable, highly potent, ATP-competitive, reversible inhibitor of both GSK-3α and β (IC₅₀ = 650 and 580 pM, respectively).
A cell-permeable, brain permeant aminopyrimidine compound that acts as a potent, ATP-competitive and reversible inhibitor of both GSK-3α and β (IC50 = 650 and 580 pM, respectively; Ki = 870 pM for human GSK-3β) and significantly affects downstream effects of GSK-3. Displays excellent selectivity over closely related Cdc2 and Erk2 (IC50 = 3.7 µM & >10 µM, respectively). Does not affect the activity of several other protein kinases studied. Acutely sensitizes glycogen synthase activity in isolated skeletal muscle from insulin-sensitive lean Zucker and insulin-resistant Zucker diabetic fatty (ZDF) rats and can also augment insulin-stimulated glucose uptake in ZDF rats. However, it does not affect the basal glucose uptake rate. Induces a significant reduction in fasting hyperglycemia in diabetic and insulin-resistant db/db mice (~30 mg/kg, s.c.). Reduces Tau phosphorylation (Ser396) in the cortex and hippocampus of postnatal rats. Also shown to activate Wnt/β-catenin signaling and direct the differentiation of human pluripotent stem cells into cardiomyocytes.
A cell-permeable, brain permeant aminopyrimidine compound that acts as a potent, ATP-competitive and reversible inhibitor of both GSK-3α and β (IC50 = 650 and 580 pM, respectively; Ki = 870 pM for human GSK-3β) and significantly affects downstream effects of GSK-3. Displays excellent selectivity over closely related Cdc2 and Erk2 (IC50 = 3.7 µM & >10 µM, respectively). Does not affect the activity of several other protein kinases studied. Acutely sensitizes glycogen synthase activity in isolated skeletal muscle from insulin-sensitive lean Zucker and insulin-resistant Zucker diabetic fatty (ZDF) rats and can also augment insulin-stimulated glucose uptake in ZDF rats. However, it does not affect the basal glucose uptake rate. Induces a significant reduction in fasting hyperglycemia in diabetic and insulin-resistant db/db mice (~30 mg/kg, s.c.). Reduces Tau phosphorylation (Ser396) in the cortex and hippocampus of postnatal rats. Also shown to activate Wnt/β-catenin signaling and direct the differentiation of human pluripotent stem cells into cardiomyocytes.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GSK-3 α & β
Target IC50: 650 and 580 pM for GSK-3&alpha
Target Ki: 870 pM for human GSK-3&beta

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Grigoryan, T., et al. 2013. Proc. Natl. Acad. Sci. USA110, 18174.
Lian, X., et al. 2012. Proc. Natl. Acad. Sci. USA109, E1848.
Selenica, M.L., et al. 2007. Br. J. Pharmacol.152, 959.
Ring, D.B., et al. 2003. Diabetes.52, 588.
Nikoulina, S.E., et al. 2002. Diabetes.51, 2190.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service