Skip to Content
Merck
All Photos(1)

Key Documents

5.31050

Sigma-Aldrich

PFKFB3 Inhibitor III, YN1

Synonym(s):

PFKFB3 Inhibitor III, YN1, 7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 6-Phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3 Inhibitor, YN1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

670 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[o]1c2c([c](c(c1)c3ccc(cc3)O)=O)ccc(c2O)O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-15-10(13(11)18)5-6-12(17)14(15)19/h1-7,16-17,19H

InChI key

BMZFZTMWBCFKSS-UHFFFAOYSA-N

General description

A cell-permeable isoflavone compound that acts as a potent and competitive inhibitor of fructose-2,6-bisphosphatase 3 (PFKFB3; IC50 = 670 nM; Ki = 240 nM). Directly docks into the Fru-6-P binding pocket of PFKFB3 and down-regulates Fru-2,6-BP levels (40% decrease in 48 h in HeLa cells) resulting in reduced glycolysis and lactate production. Shown to diminish the proliferation of HCT-116 human adenocarcinoma cells (GI50 = 8.2 µM) and inhibit HeLa cells colony formation on soft agar (~79% reduction at 25 µM). Induces apoptotic (~25 µM) and necrotic (~50 µM) cell death depending on the level of ATP depletion. Also reported to increase autophagy in HCT-116 cells as evidenced by an increase in LC3-II levels and a decrease in p62 levels
A cell-permeable isoflavone compound that acts as a potent and competitive inhibitor of fructose-2,6-bisphosphatase 3 (PFKFB3; IC50 = 670 nM; Ki = 240 nM). Directly docks into the Fru-6-P binding pocket of PFKFB3 and down-regulates Fru-2,6-BP levels (40% decrease in 48 h in HeLa cells) resulting in reduced glycolysis and lactate production. Shown to diminish the proliferation of HCT-116 human adenocarcinoma cells (GI50 = 8.2 µM) and inhibit HeLa cells colony formation on soft agar (~79% reduction at 25 µM). Induces apoptotic (~25 µM) and necrotic (~50 µM) cell death depending on the level of ATP depletion. Also reported to increase autophagy in HCT-116 cells as evidenced by an increase in LC3-II levels and a decrease in p62 levels.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Klarer, A. C., et al. 2014. Cancer Metab.2, 2.
Seo, M., et al. 2011. PLoS ONE6, e24179.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service