Skip to Content
Merck
All Photos(1)

Key Documents

5.08158

Sigma-Aldrich

TGF-β

InSolution, ≥99%, 50 mM in DMSO, RI Kinase Inhibitor II, RepSox

Synonym(s):

InSolution TGF-β RI Kinase Inhibitor II, RepSox, RepSox InSolution, E-616452 InSolution, ALK5 Inhibitor II, Transforming Growth Factor-β, Type I Receptor Kinase Inhibitor II, 2-(3-(6-Methylpyridin-2-yl)-1H-pyrazol-4-yl)-1,5-naphthyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N5
CAS Number:
Molecular Weight:
287.32
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

[nH]1ncc(c1c4nc(ccc4)C)c2nc3c(nccc3)cc2

InChI

1S/C17H13N5/c1-11-4-2-5-16(20-11)17-12(10-19-22-17)13-7-8-14-15(21-13)6-3-9-18-14/h2-10H,1H3,(H,19,22)

InChI key

LBPKYPYHDKKRFS-UHFFFAOYSA-N

General description

A cell-permeable, potent, selective, reversible, and ATP-competitive inhibitor of TGF-β type I receptor (ALK5; IC50 = 23 nM, 4 nM and 18 nM for ALK5 binding, ALK5 auto-phosphorylation, and TGF-β cellular assay in HepG2 cells, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK (IC50 >16 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ALK5
Reversible: yes
Target IC50: 23 nM, 4 nM and 18 nM for ALK5 binding, ALK5 auto-phosphorylation, and TGF-&beta

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (5 mg/348 µl) sterile-filtered solution of TGF-β RI Kinase Inhibitor II, RepSox (Cat. No. 616452) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C. Use only fresh DMSO for further dilution.

Other Notes

Ichida, J. K., et al. 2009. Cell Stem Cell5, 491.
Gellibert, F., et al. 2004. J. Med. Chem.47, 4494.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service