Skip to Content
Merck
All Photos(1)

Key Documents

5.00487

Sigma-Aldrich

Glucokinase Activator II

Synonym(s):

Glucokinase Activator II, 4-((6-(azetidine-1-carbonyl)-5-fluoropyridin-3-yl)oxy)-2-ethyl-N-(5-methylpyrazin-2-yl)-2H-indazole-6-carboxamide, Hexokinase IV Activator II, Hexokinase D Activator II, GK Activator II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22FN7O3
CAS Number:
Molecular Weight:
475.47
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

12 nM EC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable compound that reduces glucokinase Km for glucose (EC₅₀ = 120 nM & 350 nM, for human and rat enzyme). Enhances insulin secretion from INS-1 cells upon glucose exposure.
A cell-permeable indazole derivative that reduces glucokinase Km for glucose (EC50 = 120 nM and 350 nM, respectively, for human and rat enzyme) and effectively enhances insulin secretion from rat insulinima INS-1 cells upon 5 mM glucose exposure (EC50 = 170 nM). Shown to be orally available in rats (F = 69%; Cmax = 1490 ng/mL; t1/2 = 2.6 hr; AUC = 5440 ng·hr/mL; 5 mg/kg p.o.) and exhibit blood glucose lowering efficacy in OGTT (2 g/kg oral glucose challenge 1 h after single drug oral dosage) test on fasted rats (22% reduction of AUC; 250 mg/kg p.o.) in vivo.
A cell-permeable indazole derivative that reduces glucokinase Km for glucose (EC50 = 120 nM and 350 nM, respectively, for human and rat enzyme) and effectively enhances insulin secretion from rat insulinima INS-1 cells upon 5 mM glucose exposure (EC50 = 170 nM). Shown to exhibit blood glucose lowering efficacy in OGTT (2 g/kg oral glucose challenge 1 h after single drug oral dosage) test among fasted rats (22% reduction of AUC; 250 mg/kg p.o.) in vivo.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Glucokinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (=2-°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pfefferkorn, J. A., et al. 2012. Bioorg. Med. Chem. Lett.22, 7100.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service