Skip to Content
Merck
All Photos(1)

Key Documents

475956

Sigma-Aldrich

c-Myc Inhibitor

The c-Myc Inhibitor, also referenced under CAS 403811-55-2, controls the biological activity of c-Myc.

Synonym(s):

c-Myc Inhibitor, 10058-F4, ( Z,E)-5-(4-Ethylbenzylidine)-2-thioxothiazolidin-4-one, 10058-F4, (Z,E)-5-(4-Ethylbenzylidine)-2-thioxothiazolidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NOS2
CAS Number:
Molecular Weight:
249.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S1\C(=C\c2ccc(cc2)CC)\C(=O)NC1=S

InChI

1S/C12H11NOS2/c1-2-8-3-5-9(6-4-8)7-10-11(14)13-12(15)16-10/h3-7H,2H2,1H3,(H,13,14,15)/b10-7+

InChI key

SVXDHPADAXBMFB-JXMROGBWSA-N

General description

A cell-permeable thiazolidinone compound that specifically inhibits the c-Myc-Max interaction, thereby preventing the transactivation of c-Myc target gene expression. Shown to inhibit tumor cell growth in a c-Myc-dependent manner both in vitro and in vivo (64 μM using c-Myc transfected Rat1a fibroblasts).
A cell-permeable, thiazolidinone compound that specifically inhibits c-Myc-Max interaction, thereby preventing the transactivation of c-Myc targeted gene expression. Shown to inhibit tumor cell growth in a c-Myc-dependent manner both in vitro and in vivo (Effective concentration: 64 µM using c-Myc-transfected Rat1a fibroblasts).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
c-Myc
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yin, X., et al. 2003. Oncogene22, 6151.

Legal Information

Sold under license of U.S. Patent 7,026,343.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Andrea Arena et al.
Biomedicines, 10(4) (2022-04-24)
Multiple myeloma (MM) and primary effusion lymphoma (PEL) are aggressive hematological cancers, for which the search for new and more effective therapies is needed. Both cancers overexpress c-Myc and are highly dependent on this proto-oncogene for their survival. Although c-Myc
Roberta Gonnella et al.
Cancers, 15(23) (2023-12-09)
Heat shock proteins (HSPs) are highly expressed in cancer cells and represent a promising target in anti-cancer therapy. In this study, we investigated for the first time the expression of high-molecular-weight HSP110, belonging to the HSP70 family of proteins, in
Le Tran Phuc Khoa et al.
Nature communications, 15(1), 1721-1721 (2024-02-27)
Quiescence in stem cells is traditionally considered as a state of inactive dormancy or with poised potential. Naive mouse embryonic stem cells (ESCs) can enter quiescence spontaneously or upon inhibition of MYC or fatty acid oxidation, mimicking embryonic diapause in vivo.
Wei Fan et al.
eLife, 10 (2021-05-28)
Sphingolipids are important structural components of cell membranes and prominent signaling molecules controlling cell growth, differentiation, and apoptosis. Sphingolipids are particularly abundant in the brain, and defects in sphingolipid degradation are associated with several human neurodegenerative diseases. However, molecular mechanisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service