Skip to Content
Merck
All Photos(1)

Documents

474791

Sigma-Aldrich

MG-132

InSolution, ≥98%, 10 mM, reversible proteasome inhibitor

Synonym(s):

Z-Leu-Leu-Leu-al, Carbobenzoxy-L-leucyl-L-leucyl-L-leucinal, Z-LLL-CHO, Proteasome Inhibitor XI, InSolution MG-132, MG-132

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H41N3O5
CAS Number:
Molecular Weight:
475.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)OCc1ccccc1

InChI

1S/C26H41N3O5/c1-17(2)12-21(15-30)27-24(31)22(13-18(3)4)28-25(32)23(14-19(5)6)29-26(33)34-16-20-10-8-7-9-11-20/h7-11,15,17-19,21-23H,12-14,16H2,1-6H3,(H,27,31)(H,28,32)(H,29,33)/t21-,22-,23-/m0/s1

InChI key

TZYWCYJVHRLUCT-VABKMULXSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Z-Leu-Leu-Leu-al

General description

Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM). Reduces the degradation of ubiquitin-conjugated proteins in mammalian cells and permeable stains of yeast by the 26S complex without affecting its ATPase or isopeptidase activities. Activates c-Jun N-terminal kinase (JNK1) and inhibits NF-κB activation (IC50 = 3 µM).

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: yes
Target IC50: 3 µM in inhibiting NF-κB activation
Target Ki: 4 nM as proteasome inhibitor

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/210 µl) or (5mg/1050 µl) solution of MG-132 (Cat. No. 474790) in DMSO.

Other Notes

Meriin, A.B., et al. 1998. J. Biol. Chem.273, 6373.
Adams, J., and Stein, R. 1996. Ann. Rep. Med. Chem.31, 279.
Lee, D.H., and Goldberg, A.L. 1996. J. Biol. Chem.271, 27280.
Wiertz, E.J.H.J., et al. 1996. Cell84, 769.
Read, M.A., et al. 1995. Immunity2, 493.
Rock, K.L., et al. 1994. Cell78, 761.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E J Wiertz et al.
Cell, 84(5), 769-779 (1996-03-08)
Human cytomegalovirus (HCMV) down-regulates expression of MHC class I products by selective proteolysis. A single HCMV gene, US11, which encodes an endoplasmic reticulum (ER) resident type-I transmembrane glycoprotein, is sufficient to cause this effect. In US11+cells, MHC class I molecules
D H Lee et al.
The Journal of biological chemistry, 271(44), 27280-27284 (1996-11-01)
We have studied whether various agents that inhibit purified yeast and mammalian 26 S proteasome can suppress the breakdown of different classes of proteins in Saccharomyces cerevisiae. The degradation of short-lived proteins was inhibited reversibly by peptide aldehyde inhibitors of
Clarissa Braun et al.
Cell stress, 5(12), 176-182 (2021-12-18)
Programmed cell death protein 4 (PDCD4) exerts critical functions as tumor suppressor and in immune cells to regulate inflammatory processes. The phosphoinositide 3-kinase (PI3K) promotes degradation of PDCD4 via mammalian target of rapamycin complex 1 (mTORC1). However, additional pathways that
A B Meriin et al.
The Journal of biological chemistry, 273(11), 6373-6379 (1998-04-16)
Inhibition of the major cytosolic protease, proteasome, has been reported to induce programmed cell death in several cell lines, while with other lines, similar inhibition blocked apoptosis triggered by a variety of harmful treatments. To elucidate the mechanism of pro-
M A Read et al.
Immunity, 2(5), 493-506 (1995-05-01)
Multiple cell adhesion proteins are up-regulated in vascular endothelial cells in response to TNF alpha and other inflammatory cytokines. This increase in cell adhesion gene expression is thought to require the transcription factor NF-kappa B. Here, we show that peptide

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service