Skip to Content
Merck
All Photos(2)

Key Documents

434610

Sigma-Aldrich

Leukadherin-1

A furanyl thiazolidinone compound that binds CD11b αA/αI domain in a reversible manner and stabilizes αA in a high-affinity binding conformation toward known CD11b/CD18 (αMβ2; CR3; Mac-1) ligands, fibrinogen, ICAM-1, and iC3B.

Synonym(s):

Leukadherin-1, αMβ2 Agonist II, CD11b Agonist II, LA1, Mac-1 Agonist II, CR3 Agonist II, CD11b/CD18 Agonist, 4-(5-((3-Benzyl-4-oxo-2-thioxo-1,3-thiazolidin-5-ylidene)methyl)furan-2-yl)-benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15NO4S2
CAS Number:
Molecular Weight:
421.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-brown

solubility

DMSO: 5 mg/mL, yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(/C(S1)=C/C2=CC=C(C3=CC=C(C=C3)C(O)=O)O2)N(CC4=CC=CC=C4)C1=S

InChI key

AEZGRQSLKVNPCI-UNOMPAQXSA-N

General description

A furanyl thiazolidinone compound that binds CD11b αA/αI domain in a reversible manner and stabilizes αA in a high-affinity binding conformation toward known CD11b/CD18 (αMβ2; CR3; Mac-1) ligands, fibrinogen, ICAM-1, and iC3B. Reported to be superior to IMB-10 (Cat. No. 407271) in enhancing Mac-1-mediated cell adhesion (EC50 = 4 vs. >50 µM on fibrinogen surface). Effectively suppresses leukocyte recruitment in various inflammatory disease models in vivo, including thioglycolate-induced peritonitis (10 nmol/0.2 ml/mouse; i.v.), anti-GBM-induced nephritis (25 nmol/0.5 ml/mouse; i.p.), balloon-induced artery injury (50 nmol/1 ml/rat; i.m.), and zebrafish tailfin injury.CR3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Maiguel, D., et al. 2011. Sci. Signal.4, ra57.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service