Skip to Content
Merck
All Photos(1)

Documents

411492

Sigma-Aldrich

Mouse Anti-Human IgG₄, Fc Fragment Specific (HP6025)

liquid, clone HP6025, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

HP6025, monoclonal

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Purified mouse monoclonal antibody. Recognizes human IgG4 Fc fragment.
This Mouse Anti-Human IgG₄, Fc Fragment Specific (HP6025) is validated for use in Enzyme Immunoassay for the detection of Human IgG₄, Fc Fragment Specific.

Application

Enzyme Immunoassay (10 µg/ml, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Avoid more than 5 freeze/thaw cycles.

Other Notes

Antibody should be titrated for optimal results in individual systems.



Cross-reactivity by ELISA against human myeloma proteins:

Human IgG1: ≤0.1%

Human IgG2: ≤0.04%

Human IgG3: ≤0.02%

Human IgG4: 100%

Human IgA: <0.1%

Human IgE: <0.1%

Human IgM: <0.1%



Cross-reactivity by ELISA against chimeric antibodies:

Human IgG1: <0.01%

Human IgG2: <0.01%

Human IgG3: <0.01%

Human IgG4: 100%

Human IgA: <0.01%

Human IgE: <0.01%

Human IgM: <0.01%
Hamilton, R.G. 1990. Ann. Biol. Clin.48, 473.
Fasullo, F.J., et al. 1989. Clin. Chem.35, 364.
Jefferis, R., et al. 1986. Immunol. Letters10, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Houpt et al.
Infection and immunity, 62(7), 2857-2864 (1994-07-01)
Serum obtained from normal human subjects contains antibodies reactive in an enzyme-linked immunosorbent assay with the glucuronoxylomannan (GXM) of Cryptococcus neoformans. The frequency of occurrence of class-specific antibodies among normal subjects was 28% for immunoglobulin G (IgG), 98% for IgM
H Lu et al.
Infection and immunity, 61(6), 2400-2407 (1993-06-01)
The aims of this study were to determine the immunodominant antigens of Actinobacillus actinomycetemcomitans serotype b (Aab) for the different immunoglobulin (Ig) classes and subclasses and to determine the relative levels of these different Igs in serum. Seropositive early-onset periodontitis
S Sievers et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 47(8), 1057-1068 (2017-03-28)
Microarray-based component-resolved diagnostics (CRD) has become an accepted tool to detect allergen-specific IgE sensitization towards hundreds of allergens in parallel from one drop of serum. Nevertheless, specificity and sensitivity as well as a simultaneous detection of allergen-specific IgG4 , as
R G Keller et al.
Infection and immunity, 62(1), 215-220 (1994-01-01)
Previous studies found that normal human serum (NHS) contains an immunoglobulin G (IgG) antibody that mediates initiation of the classical complement pathway by nonencapsulated Cryptococcus neoformans. The present study used an enzyme-linked immunosorbent assay with whole nonencapsulated yeast cells as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service