Skip to Content
Merck
All Photos(1)

Key Documents

4060

Millipore

Inosine

A metabolite of adenosine that acts as a potent coronary vasodilator. Exhibits a positive inotropic action on heart muscle and can antagonize the ouabain-induced arrhythmias.

Synonym(s):

Inosine, Hypoxanthine Riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O5
CAS Number:
Molecular Weight:
268.23
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

Quality Level

Assay

≥99% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[n]2(c3nc[nH][c](c3nc2)=O)C1OC(C(C1O)O)CO

InChI

1S/C10H12N4O5/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18/h2-4,6-7,10,15-17H,1H2,(H,11,12,18)

InChI key

UGQMRVRMYYASKQ-UHFFFAOYSA-N

General description

A metabolite of adenosine that acts as a potent coronary vasodilator. Exhibits a positive inotropic action on heart muscle and can antagonize the ouabain-induced arrhythmias.
Correlates well with myocardial contractile performance.

Warning

Toxicity: Standard Handling (A)

Other Notes

Aviado, D.M. 1983. J. Pharmacol. 14 (suppl. 3), 47.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laureline Février et al.
Journal of environmental radioactivity, 235-236, 106645-106645 (2021-05-22)
Actinide-based mineral phases occurring in contaminated soils can be solubilized by organic chelators excreted by plants, such as citrate. Herein, the efficiency of citrate towards U and Pu extraction is compared to that of siderophores, whose primary function is the
Aiqun Lin et al.
The Journal of antibiotics, 61(4), 245-249 (2008-05-27)
Two new compounds, 4-(hydroxymethyl)-5-hydroxy-2H-pyran-2-one (1) and (5-hydroxy-2-oxo-2H-pyran-4-yl) methyl acetate (2), have been isolated from a marine-derived fungus Aspergillus flavus. Their structures were determined by spectroscopic data. Compound 1 induced the production of cAMP on GPR12 transfected CHO and HEK293 cells
Juan M Roldan-Romero et al.
International journal of cancer, 146(5), 1435-1444 (2019-07-25)
The mammalian target of rapamycin (mTOR) pathway inhibitors are key drugs for the treatment of many tumor types, however, there are no predictive biomarkers in clinical use. Here, we performed a molecular and immunohistochemical characterization of key mTOR pathway components
Hsiao-Chun Chen et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 49(5), 1813-1824 (2018-09-20)
Acupuncture involves inserting a fine needle into a specific point, often called an acupoint, thereby initiating a therapeutic effect accompanied by phenomena such as soreness, heaviness, fullness, and numbness. Acupoints are characterized as points located in deep tissues with abundant
Heejung Yang et al.
Planta medica, 79(1), 78-82 (2012-11-17)
Bioassay-guided fractionation of an 80% methanolic extract of Salix pseudo-lasiogyne twigs has resulted in the isolation of two new compounds (1-2) along with ten known ones (3-12). The new compounds were determined to be 3'-O-acetylsalicin (1) and 2',6'-O-acetylsalicortin (2) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service