Skip to Content
Merck
All Photos(1)

Key Documents

399250

Sigma-Aldrich

NG-Hydroxy-L-arginine, Monoacetate Salt

Cell permeable. A key intermediate in the biosynthesis of nitric oxide by cNOS. NOHA can be efficiently oxidized to nitric oxide and citrulline by cytochrome P450 system.

Synonym(s):

NG-Hydroxy-L-arginine, Monoacetate Salt, NOHA, AcOH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O3 · xC2H4O2
CAS Number:
Molecular Weight:
190.20 (free base basis)
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

150 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+H3][C@@H](CCC\N=C(\NO)/N)C(=O)O.[O-]C(=O)C

InChI

1S/C6H14N4O3.C2H4O2/c7-4(5(11)12)2-1-3-9-6(8)10-13;1-2(3)4/h4,13H,1-3,7H2,(H,11,12)(H3,8,9,10);1H3,(H,3,4)/t4-;/m0./s1

InChI key

VYMCYRPQICLHKC-WCCKRBBISA-N

General description

Cell permeable. A key intermediate in the biosynthesis of nitric oxide by constitutive nitric oxide synthase (cNOS). NOHA can be efficiently oxidized to nitric oxide and citrulline by cytochrome P450 system. A potent inhibitor of liver and macrophage arginase (Ki = 150 µM).
Cell-permeable. A key intermediate product in the biosynthesis of nitric oxide from arginine by constitutive nitric oxide synthase (cNOS). Formed by the NADPH-dependent hydroxylation of L-arginine. NOHA can be efficiently oxidized to nitric oxide and citrulline by cytochrome P450, providing an alternative biosynthetic route for the production of nitric oxide. A potent inhibitor of liver and macrophage arginase (Ki = 150 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
liver and macrophage arginase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Boucher, J.L., et al. 1994. Biochem. Biophys. Res. Commun. 203, 1614.
Pufahl, R.A., and Marletta, M.A. 1993. Biochem. Biophys. Res. Commun.193, 963.
Boucher, J.L., et al. 1992. Biochem. Biophys. Res. Commun.187, 880.
Stuehr, D.J., et al. 1991. J. Biol. Chem.266, 6259.
Wallace, G.C., et al. 1991. Biochem. Biophys. Res. Commun.176, 528.
Zembowicz, A., et al. 1991. Proc. Natl. Acad. Sci. USA88, 11172.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G C Wallace et al.
Biochemical and biophysical research communications, 176(1), 528-534 (1991-04-15)
This study describes the effects of N omega-hydroxy-L-arginine (NOHA) on endothelium dependent and endothelium independent relaxation of bovine pulmonary artery. These results are consistent with the hypothesis that NOHA is a biosynthetic intermediate in the production of nitric oxide from
D J Stuehr et al.
The Journal of biological chemistry, 266(10), 6259-6263 (1991-04-05)
Authentic N omega-hydroxy-L-arginine was synthesized and used to determine whether it is an intermediate in nitric oxide (.NO) synthesis from L-arginine by macrophage .NO synthase. The apparent Km (6.6 microM) and Vmax (99 nmol x min-1 x mg-1) observed with
A Zembowicz et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(24), 11172-11176 (1991-12-15)
The hypothesis was investigated that NG-hydroxy-L-arginine (L-HOArg) is an intermediate in the biosynthesis of nitric oxide (.NO) from L-arginine (L-Arg) by the constitutive .NO synthase (NOS) present in endothelial cells (ECs). When infused through a column of bovine aortic ECs
R A Pufahl et al.
Biochemical and biophysical research communications, 193(3), 963-970 (1993-06-30)
The involvement of the protoporphyrin IX heme iron of macrophage nitric oxide synthase (NOS) in the oxidation of NG-hydroxy-L-arginine (L-NHA) to nitric oxide (NO) and citrulline was investigated by carbon monoxide (CO) inhibition studies and binding difference spectroscopy. A CO:oxygen
J L Boucher et al.
Biochemical and biophysical research communications, 187(2), 880-886 (1992-09-16)
Rat liver microsomes catalyze the oxidative denitration of N omega-hydroxy-L-arginine (NOHA) by NADPH and O2 with formation of citrulline and nitrogen oxides like NO and NO2-. Besides NO2- and citrulline, whose simultaneous formation is linear for at least 20 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service