Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

370679

Sigma-Aldrich

Guanosine 3′,5′-cyclic Monophosphorothioate, β-Phenyl-1,N²-etheno-8-bromo-, Rp-Isomer, Sodium Salt

A metabolically-stable, competitive inhibitor of protein kinase G types 1α and 1β (Ki = 30 nM).

Synonym(s):

Guanosine 3′,5′-cyclic Monophosphorothioate, β-Phenyl-1,N²-etheno-8-bromo-, Rp-Isomer, Sodium Salt, Rp-8-Br-PET-cGMPS, Na, PKG Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14BrN5O6PS · Na
CAS Number:
Molecular Weight:
562.27
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[P+]1(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]3c4c(nc3Br)C(=O)[n]5c(nc(c5)c6ccccc6)N4)CO1)([S-])[O-]

InChI

1S/C18H15BrN5O6PS.Na/c19-17-21-11-14(24(17)16-12(25)13-10(29-16)7-28-31(27,32)30-13)22-18-20-9(6-23(18)15(11)26)8-4-2-1-3-5-8;/h1-6,10,12-13,16,25H,7H2,(H,20,22)(H,27,32);/q;+1/p-1/t10-,12-,13-,16-,31?;/m1./s1

General description

A metabolically-stable, competitive inhibitor of protein kinase G types 1α and 1β (Ki = 30 nM). Blocks cGMP-gated retinal type ion channels (IC50 = 25 µM) and the activation of purified PKA type II (Ki = 10 µM). More lipophilic and cell-permeable than Rp-8-pCPT-cGMPS (Cat. No. 370677). Note: 1 µmol = 0.56 mg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKG1α & 1β
Product does not compete with ATP.
Reversible: no
Target Ki: 30 nM against PKG1α & 1β

Warning

Toxicity: Standard Handling (A)

Other Notes

Wei, J.Y., et al. 1996. Biochemistry35, 16815.
Butt, E., et al. 1995. Br. J. Pharmacol. 116, 3110.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service