Skip to Content
Merck
All Photos(1)

Key Documents

341482

Sigma-Aldrich

Ferritin, Human Liver

Ferritin, Human Liver, CAS 9007-73-2, is a native, sterile-filtered ferritin that is suitable for use in immunoassays, as an immunogen, and in enzyme/radiolabeling.

Synonym(s):

Ferritin, Human Liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

human

Quality Level

description

RTECS - LK0500000

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

technique(s)

enzyme immunoassay: suitable

impurities

Antibodies to HIV1, HIV2, Hepatitis B, Hepatitis C, tested negative (by certified tests)

solubility

water: soluble

suitability

suitable for immunoassay

Protein ID accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

Research area: Cell signaling

Ferritin is a major iron storage protein. It exists in a two functionally and genetically different subunits namely, the acidic, heavy or H-chain and the basic, light or L-chain.[1] Native ferritin from human liver. Sterile-filtered.

Application

Ferritin is suitable for use in immunofluorescence assay,[2] immunoassays, as an immunogen, and in enzyme/radiolabeling.

Biochem/physiol Actions

Ferritin is a crucial protein that has multiple functions in iron metabolism and immune regulation. It helps in maintaining the iron balance.[3] H-ferritin is involved in various processes such as regulating immune function, hematopoiesis, hepatocyte apoptosis, and cell differentiation. Additionally, ferritin can act as a proinflammatory cytokine through protein kinase C-zeta (PKC-ζ) and nuclear factor kappa B (NFκB) signaling pathways in certain cells.(1) Ferritin serves as both an immune-suppressor and a signaling molecule, contributing to the regulation of lymphoid and myeloid cell proliferation.[4]

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.9% NaCl, 0.1% NaN₃, pH 7.0.

Other Notes

Addison, J.M., et al. 1983. FEBS Lett.164, 139.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ferritin, iron homeostasis, and oxidative damage
Arosio P and Levi S
Free radical biology & medicine, 33(4), 457-463 (2002)
Ferritin functions as a proinflammatory cytokine via iron-independent protein kinase C zeta/nuclear factor kappaB-regulated signaling in rat hepatic stellate cells
Ruddell RG, et al.
Journal of Separation Science, 49(3), 887-900 (2009)
Ferritin functions as a proinflammatory cytokine via iron-independent protein kinase C zeta/nuclear factor kappaB-regulated signaling in rat hepatic stellate cells
Ruddell RG, et al.
Hepatology, 49(3), 887-900 (2009)
New functions for an iron storage protein: the role of ferritin in immunity and autoimmunity
Recalcati S, et al.
Journal of Autoimmunity, 30(1-2), 84-89 (2008)
Ferritin, iron homeostasis, and oxidative damage
Arosio P, et al.
Journal of Separation Science, 33(4), 457-463 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service