Skip to Content
Merck
All Photos(1)

Key Documents

324725

Sigma-Aldrich

Endoglycosidase F1, Elizabethkingia meningosepticum, Recombinant, E. coli

Endoglycosidase F1, Elizabethkingia meningosepticum, Recombinant, E. coli cleaves asparagine-linked or free oligomannose and hybrid. Suitable for deglycosylation of native proteins.

Synonym(s):

Endo-β-N-acetylglucosaminidase F1, Endo F1

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

form

liquid

specific activity

≥16 units/mg protein
≥17 units/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

Proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, Elizabethkingia meningosepticum endoglycosidase F1 expressed in E. coli. Cleaves asparagine-linked or free oligomannose and hybrid, but not complex oligosaccharides. Core fucosylation reduces activity by 50 fold. Endo F1 will hydrolyze sulfate containing high mannose chains. It cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine. Less sensitive to protein conformation than N-Glycosidase F (Cat. No. 362185) and therefore is more suitable for deglycosylation of native proteins.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release N-linked oligosaccharides from 1.0 µmol denatured ribonuclease B per min at 37°C, pH 5.5.

Physical form

In 20 mM Tris-HCl, pH 7.5.

Other Notes

Tarentino, A.L., and Plummer, T.H. 1994. Methods Enzymol. 230, 44.
Tarentino, A.L., et al. 1992. J. Biol. Chem. 267, 3868.
Trimble, R.B., and Tarentino, A.L. 1991. J. Biol. Chem. 266, 1646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Tarentino et al.
The Journal of biological chemistry, 267(6), 3868-3872 (1992-03-06)
A full-length insert for the endo-beta-N-acetylglucosaminidase (Endo) F1 gene was located on a 2,200-base pair EcoRI fragment of genomic DNA and cloned into the plasmid vector Bluescript. Transformed Escherichia coli cells expressed Endo F1 activity very well, but the enzyme
R B Trimble et al.
The Journal of biological chemistry, 266(3), 1646-1651 (1991-01-25)
Flavobacterium meningosepticum endo-beta-acetyl-glucosaminidase F preparations have been resolved by hydrophobic interaction chromatography on TSK-butyl resin into at least three activities designated endo F1, endo F2 and endo F3 each with a unique substrate specificity. The 32-kDa endo F1 protein is
Thapakorn Jaroentomeechai et al.
Nature communications, 13(1), 6325-6325 (2022-10-25)
The ability to reconstitute natural glycosylation pathways or prototype entirely new ones from scratch is hampered by the limited availability of functional glycoenzymes, many of which are membrane proteins that fail to express in heterologous hosts. Here, we describe a
Enzymatic deglycosylation of asparagine-linked glycans: purification, properties, and specificity of oligosaccharide-cleaving enzymes from Flavobacterium meningosepticum.
A L Tarentino et al.
Methods in enzymology, 230, 44-57 (1994-01-01)

Articles

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service