Skip to Content
Merck
All Photos(1)

Key Documents

234409

Sigma-Aldrich

Complement C9-Depleted Serum, Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25
form:
liquid
shipped in:
dry ice

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice

storage temp.

−70°C

General description

Human serum depleted of complement component C9 by affinity chromatography. C9-depleted serum, sterile-filtered through a 0.22 µm filter. No C9 detected by Ouchterlony using various dilutions of goat anti-human C9 antibody.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

Classical Pathway Activity: One C9H50 unit measured by classical pathway activation is defined as the amount of C9 required to yield 50% lysis of 3 x 10⁷EA when incubated in the presence of the recommended volume of C9-Dpl for 30 min at 37°C in a total reaction volume of 500 µl GVB++.CH50 Functional Activity upon Reconstitution: One CH50 unit is defined as the input of C9-Dpl, reconstituted with C9, or NHS Complement Standard yielding 50% lysis of 1 x 10⁸ EA when incubated for 60 min at 37°C in a total reaction volume of 1.5 ml GVB++.Alternative Pathway Activity: One unit of whole alternative pathway activity (APH50) is defined as the input of C9-Dpl, reconstituted with C9, or NHS Complement Standard yielding 50% lysis of 1.5 x 10⁷ rabbit erythrocytes when incubated for 30 min at 37°C in a total reaction volume of 75 µl GVB° containing a final Mg-EGTA concentration of 13.3 mM.

Physical form

In 150 mM NaCl, 50 mM sodium phosphate, pH 7.2 ± 0.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HCV, HIV-I, and HIV-II.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service