Skip to Content
Merck
All Photos(1)

Documents

220488

Sigma-Aldrich

Chk2 Inhibitor, PV1019

The Chk2 Inhibitor, PV1019 controls the biological activity of Chk2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Chk2 Inhibitor, PV1019, NSC 744039

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17N7O3
Molecular Weight:
379.37
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable nitroindolo-guanidinohydrazone compound that acts as a potent, ATP-competitive, and highly selective Chk2 inhibitor (IC50 = 138 and 570 nM against the phosphorylation of histone H1 by Chk2 in the presence of 10 µM and 1 mM of ATP, respectively), while exhibiting much reduced or no activity against a panel of 52 other cellular kinases, including Chk1 (IC50 = 15.73 and 55 µM against substrate phosphorylation and Chk1 autophosphorylation with 10 µM ATP). Shown to effectively suppress radiation-induced phosphorylation of Chk2 (on Ser516) and its downstream substrate Cdc25c (on Ser216) in MCF7 cultures (IC50<10 µM) and reduce the sensitivity of wild-type murine thymocytes to radiation-induced apoptosis to the level seen in Chk2-/- cultures.
A cell-permeable nitroindolo-guanidinohydrazone compound that acts as a potent, ATP-competitive, and highly selective Chk2 inhibitor (IC50 = 138 and 570 nM against the phosphorylation of histone H1 by Chk2 in the presence of 10 µM and 1 mM of ATP, respectively), while exhibiting much reduced or no activity against a panel of 52 other cellular kinases, including Chk1 (IC50 = 15.73 and 55 µM against substrate phosphorylation and Chk1 autophosphorylation with 10 µM ATP). Shown to effectively suppress radiation-induced phosphorylation of Chk2 (on Ser516) and its downstream substrate Cdc25c (on Ser216) in MCF7 cultures (IC50<10 µM) and reduce the sensitivity of wild-type murine thymocytes to radiation-induced apoptosis to the level seen in Chk2-/- cultures.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliqout and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jobson, A.G., et al. 2009. J. Pharmacol. Exp. Ther.331, 816.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho Tsoi et al.
International journal of molecular sciences, 23(20) (2022-10-28)
Breast cancer is a heterogeneous disease. Tamoxifen is frequently used to treat ER-positive breast cancer. Our team has identified a novel splice variant of NCOR2, BQ323636.1 (BQ), that mediates tamoxifen resistance. However, the upstream factors that modulate BQ expression are
Gwenola Manic et al.
Cell death and differentiation, 28(7), 2060-2082 (2021-02-04)
Cancer stem cells (CSCs) are tumor subpopulations driving disease development, progression, relapse and therapy resistance, and their targeting ensures tumor eradication. CSCs display heterogeneous replication stress (RS), but the functionality/relevance of the RS response (RSR) centered on the ATR-CHK1 axis
Brandoch Cook et al.
The Journal of biological chemistry, 296, 100696-100696 (2021-04-26)
Bone fractures are common impact injuries typically resolved through natural processes of osteogenic regeneration and bone remodeling, restoring the biological and mechanical function. However, dysfunctionality in bone healing and repair often arises in the context of aging-related chronic disorders, such
Yandong Yin et al.
Molecular cell, 81(20), 4243-4257 (2021-09-03)
Mammalian cells use diverse pathways to prevent deleterious consequences during DNA replication, yet the mechanism by which cells survey individual replisomes to detect spontaneous replication impediments at the basal level, and their accumulation during replication stress, remain undefined. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service