Skip to Content
Merck
All Photos(1)

Key Documents

203291

Sigma-Aldrich

Bisindolylmaleimide I, Hydrochloride

An enhanced water-soluble form of Bisindolylmaleimide I.

Synonym(s):

Bisindolylmaleimide I, Hydrochloride, 2-[1-(3-Dimethylaminopropyl)-1H-indol-3-yl]-3-(1H-indol-3-yl)maleimide, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N4O2 · xHCl
CAS Number:
Molecular Weight:
412.48 (free base basis)
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange-red

solubility

water: 10 mg/mL
DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl.N1C(=O)C(=C(C1=O)c4c5c([nH]c4)cccc5)c2c3c([n](c2)CCCN(C)C)cccc3

InChI

1S/C25H24N4O2.ClH/c1-28(2)12-7-13-29-15-19(17-9-4-6-11-21(17)29)23-22(24(30)27-25(23)31)18-14-26-20-10-5-3-8-16(18)20;/h3-6,8-11,14-15,26H,7,12-13H2,1-2H3,(H,27,30,31);1H

InChI key

XRAMWNCMYJHGGH-UHFFFAOYSA-N

General description

A reversible and ATP-competitive inhibitor of protein kinase C (Ki = 10 nM). Inhibits protein kinase A at much higher concentrations (Ki = 2 µM). An enhanced water soluble form of Bisindolylmaleimide I (Cat. No. 203290).
An enhanced water-soluble form of Bisindolylmaleimide I (Cat. No. 203290). A reversible and ATP-competitive inhibitor of protein kinase C (Ki = 10 nM). May inhibit protein kinase A at much higher concentrations (Ki = 2 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Protein kinase C
Product competes with ATP.
Reversible: yes
Target Ki: 10 nM against protein kinase C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Le Panse, R., et al. 1994. Mol. Pharmacol.46, 445.
Kuchera, S., et al. 1993. Agents Actions 39, C169.
Wilkinson, S.E., et al. 1993. Biochem. J.294, 335.
Toullec, D., et al. 1991. J. Biol. Chem. 266, 15771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Christopher et al.
The Journal of physiology, 515 ( Pt 2), 511-522 (1999-03-02)
1. During development, embryos of the pulmonate gastropod, Helisoma trivolvis, undergo a rotation behaviour due to the co-ordinated beating of three bands of ciliated epithelial cells. This behaviour is in part mediated by the neurotransmitter serotonin (5-HT) released from a
Stephen R Reeves et al.
Experimental physiology, 92(6), 1057-1066 (2007-08-07)
Protein kinase C (PKC) is a broadly expressed and critically important signalling protein with a wide range of functional roles, including central components of respiratory control. For example, systemic and targeted administration of PKC inhibitors within the nucleus of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service