Skip to Content
Merck
All Photos(1)

Key Documents

20-176

Sigma-Aldrich

100X GTPγS, 10mM

Synonym(s):

GTP gamma S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity binding assay: suitable (G-protein)

shipped in

dry ice

General description

Guanosine5′-O-(3-thio)triphosphate (GTPγS) is a slowly hydrolyzable and non-hydrolyzableGTP analog. It acts as a G-protein-activating nucleotide. GTPγS canstimulate G protein-coupled receptor (GPCR) activity.

Application

GTP gamma S for use as a positive loading control in the Activation Assay Kits
Research Category
All

Physical form

sterile water

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive and selective liquid-chromatographic assay of fluoxetine and norfluoxetine in plasma with fluorescence detection after precolumn derivatization.
R F Suckow,M F Zhang,T B Cooper
Clinical Chemistry null
Considerations when measuring urinary albumin: precision, substances that may interfere, and conditions for sample storage.
M L MacNeil,P W Mueller,S P Caudill,K K Steinberg
Clinical Chemistry null
Analysis of 8-methoxypsoralen by high-performance liquid chromatography.
C H Ketchum,C A Robinson,S T Huang
Clinical Chemistry null
L Hagen et al.
Clinical chemistry, 39(1), 134-138 (1993-01-01)
High-performance ion chromatography (HPIC) has been widely used for oxalate analysis and, more recently, for glycolate analysis. We describe a procedure for sample preparation in which the plasma ultrafiltrate is acidified during harvesting with a cation-exchange resin, and the chloride
Sodium iron EDTA [NaFe(III)EDTA] as a food fortificant: the effect on the absorption and retention of zinc and calcium in women.
L Davidsson,P Kastenmayer,R F Hurrell
American Journal of Clinical Nutrition null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service